Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00781102_s1
          See other ZFP280B GE Assays ›
          Gene Symbol:
          Zfp280b
          Gene Name:
          zinc finger protein 280B
          Gene Aliases:
          D10Jhu82e, Suhw2
          Chromosome Location:
          Chr.10: 76032401 - 76042970 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00781102_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp280b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177475.3 NP_803426.2 2 4765 100
          GenBank mRNA AK031331.1 - 2 4665 100
          AK079824.1 - 1 2432 100
          AK144644.1 - 2 4722 99
          AK144736.1 - 2 4721 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp280b

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          biological_process
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27259912 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27259913 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486068 AK031331.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P486068 AK031331.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81345 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116383 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138978 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125013 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486068 AK031331.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P486068 AK031331.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27259901 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125013 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138978 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116383 - -
          Whole Mouse Genome G4122A A_51_P486068 AK031331.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P486068 AK031331.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116383 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125013 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138978 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08402352 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08402364 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08402358 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27259912 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27259913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27259946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08530037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08794009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08379600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08530027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08530023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27259948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08402364 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364131 - -
          Affymetrix HT MG-430 PM Array Plate 1460564_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10364130 - -
          Mouse Genome 430 2.0 Array 1460564_PM_at - -
          Mouse Genome 430 2.0 Array 1460564_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64453_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 64453_at - -
          Affymetrix Mouse Clariom S Array PSR1000005245.mm.1 NM_177475.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005245.mm.1 NM_177475.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005245.mm.1 TC1000000733.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005244.mm.1 NM_177475.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005244.mm.1 NM_177475.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005244.mm.1 TC1000000733.mm.1 MTA TC
          Mouse Expression Set 430 1460564_at - -
          Mouse Expression Set 430 1460564_PM_at - -
          Murine Genome U74v2 Set 166566_i_at - -
          Murine Genome U74v2 Set 166566_PM_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1460564_at - -
          Affymetrix HT Mouse Genome 430B Array 1460564_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6180619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2744924 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2744924 - -
          mouse-6 v1.1 (Illumina) scl38754.2_721-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38754.2_721-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2804702 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2804702 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl38754.2_721-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0