Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00781309_s1
          See other 9830107B12RIK GE Assays ›
          Gene Symbol:
          9830107B12Rik
          Gene Name:
          RIKEN cDNA 9830107B12 gene
          Gene Aliases:
          Trem5
          Chromosome Location:
          Chr.17: 48098750 - 48146307 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00781309_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9830107B12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177896.1 NP_001171367.1 4 3077 126
          NM_001177897.1 NP_001171368.1 4 2930 126
          NM_177824.4 NP_808492.1 3 2986 126
          GenBank mRNA AK079398.1 - 3 2947 126

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 9830107B12Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678206 AK079398.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678206 NM_177824.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678206 NM_001177897.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678206 NM_001177896.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678206 AK079398.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678206 NM_177824.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678206 NM_001177897.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P678206 NM_001177896.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030996 EF646801.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030996 AK154301.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030996 AK169970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030996 AK079398.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030996 AK137568.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 574338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 273458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 31512 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155101 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158955 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678206 AK079398.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678206 NM_177824.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678206 NM_001177897.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678206 NM_001177896.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678206 AK079398.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678206 NM_177824.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678206 NM_001177897.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P678206 NM_001177896.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030996 EF646801.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030996 AK154301.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030996 AK169970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030996 AK079398.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2030996 AK137568.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08794192 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03327697 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155101 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03327700 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158955 - -
          Whole Mouse Genome G4122A A_52_P678206 AK079398.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678206 NM_177824.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678206 NM_001177897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678206 NM_001177896.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678206 AK079398.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678206 NM_177824.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678206 NM_001177897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P678206 NM_001177896.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102131 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101402 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155101 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158955 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03327700 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13551096 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13399447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31215998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31216030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13509548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13476854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13466428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13399448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13476861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31216152 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10356620 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451645 - -
          Affymetrix Mouse Gene 1.1 ST Array 10356601 - -
          Affymetrix Mouse Gene 1.1 ST Array 10451641 - -
          Affymetrix Mouse Clariom S Array PSR1700020996.mm.1 NM_001177896.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020996.mm.1 NM_001177896.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020996.mm.1 NM_001177897.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020996.mm.1 NM_001177897.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020996.mm.1 NM_177824.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020996.mm.1 NM_177824.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020996.mm.1 TC1700002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020994.mm.1 NM_001177896.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020994.mm.1 NM_001177896.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020994.mm.1 NM_001177897.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020994.mm.1 NM_001177897.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020994.mm.1 NM_177824.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020994.mm.1 NM_177824.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020994.mm.1 TC1700002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020995.mm.1 NM_001177896.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020995.mm.1 NM_001177896.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020995.mm.1 NM_001177897.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020995.mm.1 NM_001177897.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020995.mm.1 TC1700002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020998.mm.1 NM_001177896.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020998.mm.1 NM_001177896.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020998.mm.1 NM_001177897.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020998.mm.1 NM_001177897.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020998.mm.1 NM_177824.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020998.mm.1 NM_177824.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020998.mm.1 TC1700002223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700020997.mm.1 NM_001177896.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020997.mm.1 NM_001177896.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020997.mm.1 NM_177824.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020997.mm.1 NM_177824.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700020997.mm.1 TC1700002223.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 513191 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6760139 - -
          Illumina mouse-6 v1.0 expression beadchip 6760139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239752 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239752 - -
          mouse-6 v1.1 (Illumina) scl0328829.1_249-S - -
          Illumina Sentrix mouse-6.v1 bead array 6760139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0328829.1_249-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239752 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239752 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0