Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00781429_s1
          See other PRAMEL7 GE Assays ›
          Gene Symbol:
          Pramel7
          Gene Name:
          preferentially expressed antigen in melanoma like 7
          Gene Aliases:
          AA675346, C81180
          Chromosome Location:
          Chr.2: 87489088 - 87492418 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00781429_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pramel7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178250.2 NP_839981.1 3 1659 137
          GenBank mRNA AF490343.1 - 3 1659 137
          BC052825.1 - 3 1670 30030731 137

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pramel7

          Gene Ontology Categories:

          Function(s) Process(es)

          stem cell population maintenance
          regulation of ERK1 and ERK2 cascade
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078660 AF490343.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078660 BC052825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078660 NM_178250.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101815 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131856 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078660 AF490343.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078660 BC052825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078660 NM_178250.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406154 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131856 - -
          Whole Mouse Genome G4122A A_52_P200451 AF490343.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P200451 BC052825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P200451 NM_178250.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116862 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01295287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01295289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01295284 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01727248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01981827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01295284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01184635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01295289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01184638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02111410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02034958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01931759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01184637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01295287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01289058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01489898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01184646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01230991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01489894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02034961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02096829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02254407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406131 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484672 - -
          Affymetrix HT MG-430 PM Array Plate 1439809_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439810_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10484669 - -
          Mouse Genome 430 2.0 Array 1439809_at - -
          Mouse Genome 430 2.0 Array 1439810_PM_s_at - -
          Mouse Genome 430 2.0 Array 1439809_PM_at - -
          Mouse Genome 430 2.0 Array 1439810_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 347712_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 347712_at - -
          Mouse Expression Set 430 1439810_s_at - -
          Mouse Expression Set 430 1439810_PM_s_at - -
          Mouse Expression Set 430 1439809_at - -
          Mouse Expression Set 430 1439809_PM_at - -
          Murine Genome U74v2 Set 93245_PM_at - -
          Murine Genome U74v2 Set 93245_at - -
          Mouse Genome 430A 2.0 Array 1439809_at - -
          Mouse Genome 430A 2.0 Array 1439810_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1439810_s_at - -
          Mouse Genome 430A 2.0 Array 1439809_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439810_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1439809_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1439810_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1439809_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 342598 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6860347 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860347 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256946 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256946 - -
          mouse-6 v1.1 (Illumina) scl19043.4.1_203-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19043.4.1_203-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860347 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256946 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256946 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline