Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00781525_s1
          See other ZFP764 GE Assays ›
          Gene Symbol:
          Zfp764
          Gene Name:
          zinc finger protein 764
          Gene Aliases:
          8030466O12Rik
          Chromosome Location:
          Chr.7: 127382263 - 127406835 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00781525_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp764
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167832.1 NP_001161304.1 3 1628 147
          NM_146203.4 NP_666315.3 3 1665 147
          GenBank mRNA AK042873.1 - 3 1666 147
          AK053795.1 - 3 1590 147
          BC026397.1 - 3 1550 4207307 147

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp764

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505509 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 BC026397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 AK042873.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 AK053795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 NM_001167832.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 NM_146203.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 BC026397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 AK042873.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 AK053795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 NM_001167832.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P317673 NM_146203.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 BC026397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 AK042873.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 AK053795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 NM_001167832.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 NM_146203.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 BC026397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 AK042873.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 AK053795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 NM_001167832.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P317673 NM_146203.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505515 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06361556 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06361564 - -
          Whole Mouse Genome G4122A A_51_P317673 BC026397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317673 AK042873.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317673 AK053795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317673 NM_001167832.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317673 NM_146203.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317673 BC026397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317673 AK042873.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317673 AK053795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317673 NM_001167832.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P317673 NM_146203.4 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05912002 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06437730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06603820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06248883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06371329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505504 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568246 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568244 - -
          Affymetrix HT MG-430 PM Array Plate 1424916_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568243 - -
          Mouse Genome 430 2.0 Array 1424916_PM_x_at - -
          Mouse Genome 430 2.0 Array 1424916_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233893_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233893_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700036338.mm.1 NM_001167832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036338.mm.1 NM_001167832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036338.mm.1 NM_146203.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036338.mm.1 NM_146203.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036338.mm.1 TC0700004326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036335.mm.1 NM_146203.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036335.mm.1 NM_146203.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036335.mm.1 TC0700004326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036333.mm.1 NM_001167832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036333.mm.1 NM_001167832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036333.mm.1 NM_146203.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036333.mm.1 NM_146203.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036333.mm.1 TC0700004326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036334.mm.1 NM_001167832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036334.mm.1 NM_001167832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036334.mm.1 NM_146203.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036334.mm.1 NM_146203.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036334.mm.1 TC0700004326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036336.mm.1 NM_001167832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036336.mm.1 NM_001167832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036336.mm.1 NM_146203.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036336.mm.1 NM_146203.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036336.mm.1 TC0700004326.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036337.mm.1 NM_001167832.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036337.mm.1 NM_001167832.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036337.mm.1 NM_146203.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036337.mm.1 NM_146203.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036337.mm.1 TC0700004326.mm.1 MTA TC
          Mouse Expression Set 430 1424916_PM_x_at - -
          Mouse Expression Set 430 1451571_s_at - -
          Mouse Expression Set 430 1424916_x_at - -
          Mouse Expression Set 430 1451571_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1424916_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1424916_x_at - -
          Mouse Genome 430A 2.0 Array 1451571_s_at - -
          Mouse Genome 430A 2.0 Array 1451571_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424916_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1424916_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 592055 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3850402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850402 - -
          mouse-6 v1.1 (Illumina) scl30646.3_608-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850402 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30646.3_608-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2960774 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225405 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2960774 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl30646.3_608-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline