Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00781581_s1
          See other HIST1H2AE GE Assays ›
          Gene Symbol:
          Hist1h2ae
          Gene Name:
          histone cluster 1, H2ae
          Gene Aliases:
          1190022L06
          Chromosome Location:
          Chr.13: 23570663 - 23571220 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00781581_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h2ae
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK028026.1 - 1 652 123

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          histone

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882280 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882281 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882282 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286460 AK028026.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286460 NM_178187.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286460 AK028026.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286460 NM_178187.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286460 AK028026.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286460 NM_178187.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286460 AK028026.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286460 NM_178187.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286460 AK028026.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286460 NM_178187.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286460 AK028026.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P286460 NM_178187.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882280 - -
          Whole Mouse Genome G4122A A_51_P286460 AK028026.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286460 NM_178187.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286460 AK028026.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286460 NM_178187.3 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10525880 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10525889 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10525888 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882282 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882281 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10525870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882275 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10408206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408207 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404047 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408209 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404045 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408204 - -
          Mouse Expression Set 430 1418367_x_at - -
          Mouse Expression Set 430 1418366_at - -
          Mouse Expression Set 430 1418366_PM_at - -
          Mouse Expression Set 430 1438009_PM_at - -
          Mouse Expression Set 430 1438009_at - -
          Mouse Expression Set 430 1418367_PM_x_at - -
          Murine Genome U74v2 Set 94805_f_at - -
          Murine Genome U74v2 Set 94805_PM_f_at - -
          Murine Genome U74v2 Set 167053_PM_at - -
          Murine Genome U74v2 Set 167053_at - -
          Mouse Genome 430A 2.0 Array 1418366_PM_at - -
          Mouse Genome 430A 2.0 Array 1418367_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1418366_at - -
          Mouse Genome 430A 2.0 Array 1438009_at - -
          Mouse Genome 430A 2.0 Array 1438009_PM_at - -
          Mouse Genome 430A 2.0 Array 1418367_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 362206 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103190066 - -
          Illumina mouse-6 v1.0 expression beadchip 103190066 - -
          Illumina mouse-6 v1.0 expression beadchip 4920672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004920672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2751102 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2751102 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221597 - -
          mouse-6 v1.1 (Illumina) scl44212.1.1_304-S - -
          mouse-6 v1.1 (Illumina) GI_31982205-S - -
          Illumina Sentrix mouse-6.v1 bead array 103190066 - -
          Illumina Sentrix mouse-6.v1 bead array 4920672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44212.1.1_304-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4920672 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103190066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2751102 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2751102 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline