Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00781827_s1
          See other VMN1R73 GE Assays ›
          Gene Symbol:
          Vmn1r73
          Gene Name:
          vomeronasal 1 receptor 73
          Gene Aliases:
          V1rg2
          Chromosome Location:
          Chr.7: 11756257 - 11757168 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00781827_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134203.1 NP_598964.1 1 453 150

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r73

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977169 NM_134203.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138135 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977169 NM_134203.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P130233 NM_134203.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138135 - -
          Whole Mouse Genome G4122A A_51_P130233 NM_134203.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123793 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06572893 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06039657 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06330620 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06039658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06039661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06458130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06527354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05995457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05944749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06039651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06135723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969493 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549983 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549982 - -
          Affymetrix Mouse Clariom S Array PSR0700001029.mm.1 TC0700000118.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001029.mm.1 NM_134203.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001029.mm.1 NM_134203.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 632496 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4670497 - -
          Illumina mouse-6 v1.0 expression beadchip 4670497 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670497 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2467955 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2467955 - -
          mouse-6 v1.1 (Illumina) scl9724.1.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670497 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9724.1.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670497 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2467955 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2467955 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline