Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00782119_s1
          See other CETN4 GE Assays ›
          Gene Symbol:
          Cetn4
          Gene Name:
          centrin 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 37308627 - 37312446 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00782119_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cetn4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145825.2 NP_665824.1 5 749 132
          GenBank mRNA AF362367.1 - 5 749 132
          AK078275.1 - 4 2664 132
          AK090064.1 - 607 132
          AK166515.1 - 3 527 132
          BC038154.1 - 4 640 132
          BC060991.1 - 5 701 6742836 132

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cetn4

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          calcium ion binding
          G-protein beta/gamma-subunit complex binding
          heterotrimeric G-protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22062863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22062859 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22062861 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22062862 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029294 AK131808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029294 BC038154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029294 BC060991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029294 NM_145825.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029294 AF362367.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980539 AK090064.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980539 AK166515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980539 BC038154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980539 BC060991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1980539 BC087905.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 110477 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41709 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128674 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029294 AK131808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029294 BC038154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029294 BC060991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029294 NM_145825.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029294 AF362367.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980539 AK090064.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980539 AK166515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980539 BC038154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980539 BC060991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1980539 BC087905.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108990 AK131808.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108990 AK166515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108990 BC038154.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108990 BC060991.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P108990 NM_145825.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22062875 - -
          Whole Mouse Genome G4122A A_51_P108990 AK131808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108990 AK166515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108990 BC038154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108990 BC060991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P108990 NM_145825.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135186 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02433700 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02433701 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22062862 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22062863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22062859 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22062861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22062774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02909734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03054591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02860279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02479572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02909727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03054590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02380513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02433710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02433697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02433700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22062837 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10345075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10345076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497895 - -
          Affymetrix HT MG-430 PM Array Plate 1445511_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436617_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10497894 - -
          Affymetrix Mouse Gene 1.1 ST Array 10345074 - -
          Mouse Genome 430 2.0 Array 1445511_PM_at - -
          Mouse Genome 430 2.0 Array 1436617_at - -
          Mouse Genome 430 2.0 Array 1445511_at - -
          Mouse Genome 430 2.0 Array 1436617_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 207175_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 207175_at - -
          Affymetrix Mouse Clariom S Array PSR0300014466.mm.1 NM_145825.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014466.mm.1 NM_145825.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014466.mm.1 TC0300001869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300014467.mm.1 NM_145825.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014467.mm.1 NM_145825.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300014467.mm.1 TC0300001869.mm.1 MTA TC
          Mouse Expression Set 430 1436617_PM_at - -
          Mouse Expression Set 430 1445511_at - -
          Mouse Expression Set 430 1436617_at - -
          Mouse Expression Set 430 1445511_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445511_at - -
          Affymetrix HT Mouse Genome 430B Array 1436617_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445511_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436617_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 652160 - -
          ABI Mouse Genome Survey Microarray 685710 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_3032453 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3105936 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3032453 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3105936 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline