Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00783118_s1
          See other GCSH GE Assays ›
          Gene Symbol:
          Gcsh
          Gene Name:
          glycine cleavage system protein H (aminomethyl carrier)
          Gene Aliases:
          1100001L02Rik, 5730591C18Rik
          Chromosome Location:
          Chr.8: 116981828 - 116993449 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00783118_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gcsh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026572.3 NP_080848.1 5 1279 84
          GenBank mRNA AK019983.1 - 5 1280 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gcsh

          Gene Ontology Categories:

          Function(s) Process(es)

          glycine decarboxylation via glycine cleavage system
          methylation
          aminomethyltransferase activity
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200118 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200119 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200116 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200120 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200115 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018549 AK009543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018549 AK019983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018549 AK050373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018549 NM_026572.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018549 AK003189.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109800 AK019983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109800 NM_026572.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102250 AK019983.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102250 NM_026572.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884294 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 291125 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 115423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98804 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103078 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114472 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018549 AK009543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018549 AK019983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018549 AK050373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018549 NM_026572.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018549 AK003189.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109800 AK019983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109800 NM_026572.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102250 AK019983.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102250 NM_026572.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26200106 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114472 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103078 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01838458 - -
          Whole Mouse Genome G4122A A_52_P390625 AK019983.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P390625 AK050373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P390625 BC014745.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P390625 NM_026572.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P390625 AK003788.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109800 AK019983.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109800 NM_026572.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103078 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114472 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07346198 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07318789 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07318777 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07318774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07346195 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200118 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200116 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200120 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200115 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26200119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26200060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07429886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07318773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06779984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06931288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07029479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06743069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06726465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07318775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06816545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07318790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07318783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07429884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07346198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26200119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06726462 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10582023 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582022 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582024 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582021 - -
          Affymetrix HT MG-430 PM Array Plate 1432164_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10582020 - -
          Mouse Genome 430 2.0 Array 1432164_PM_a_at - -
          Mouse Genome 430 2.0 Array 1432164_a_at - -
          Murine Genome U74v2 Set 160385_at - -
          Murine Genome U74v2 Set 160385_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1432164_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432164_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 530464 - -
          ABI Mouse Genome Survey Microarray 674460 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4560040 - -
          Illumina mouse-6 v1.0 expression beadchip 4560040 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004560040 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2696216 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2696216 - -
          mouse-6 v1.1 (Illumina) scl34278.8_602-S - -
          Illumina Sentrix mouse-6.v1 bead array 4560040 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34278.8_602-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4560040 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2696216 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2696216 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline