Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00783163_s1
          See other LCE1E GE Assays ›
          Gene Symbol:
          Lce1e
          Gene Name:
          late cornified envelope 1E
          Gene Aliases:
          1110031B11Rik, AI507275
          Chromosome Location:
          Chr.3: 92707400 - 92709074 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00783163_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lce1e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026811.2 NP_081087.1 2 628 74
          GenBank mRNA AK003998.1 - 2 629 74
          BC059085.1 - 1 587 74
          BC099588.1 - 1 586 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lce1e

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314465 BC059085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314465 BC099588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314465 AK003998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314465 NM_026811.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314465 BC059085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314465 BC099588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314465 AK003998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P314465 NM_026811.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55380 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314465 BC059085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314465 BC099588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314465 AK003998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314465 NM_026811.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314465 BC059085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314465 BC099588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314465 AK003998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P314465 NM_026811.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332749 - -
          Whole Mouse Genome G4122A A_51_P314465 BC059085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314465 BC099588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314465 AK003998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314465 NM_026811.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314465 BC059085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314465 BC099588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314465 AK003998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P314465 NM_026811.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130114 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04989815 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02329818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02329823 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02329822 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02329819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02313813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03152526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02956193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02329819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03152521 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499925 - -
          Affymetrix Mouse Gene 1.1 ST Array 10499924 - -
          Affymetrix Mouse Clariom S Array PSR0300018626.mm.1 NM_026811.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018626.mm.1 NM_026811.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018626.mm.1 TC0300002405.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018627.mm.1 NM_026811.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018627.mm.1 NM_026811.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018627.mm.1 TC0300002405.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 645656 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6760717 - -
          Illumina mouse-6 v1.0 expression beadchip 510021 - -
          Illumina mouse-6 v1.0 expression beadchip 6760717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006760717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510021 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699586 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658001 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658001 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2699586 - -
          mouse-6 v1.1 (Illumina) scl0068694.1_315-S - -
          mouse-6 v1.1 (Illumina) scl068694.4_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 510021 - -
          Illumina Sentrix mouse-6.v1 bead array 6760717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068694.4_24-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0068694.1_315-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510021 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6760717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2658001 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258290 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2658001 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258290 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline