Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00783361_s1
          See other LYSMD1 GE Assays ›
          Gene Symbol:
          Lysmd1
          Gene Name:
          LysM, putative peptidoglycan-binding, domain containing 1
          Gene Aliases:
          2610022K04Rik, AI415311
          Chromosome Location:
          Chr.3: 95134088 - 95139525 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00783361_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lysmd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153121.2 NP_694761.1 3 2356 84
          GenBank mRNA AK011517.1 - 3 2269 84
          AK154180.1 - 3 2357 84
          BC024838.1 - 3 2334 5360726 84
          BC100548.1 - 3 2059 5717642 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lysmd1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859303 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151860 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974617 BC024838.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974617 BC100548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974617 AK154180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974617 NM_153121.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974617 AK011517.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124341 BC024838.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124341 BC100548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124341 AK154180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124341 NM_153121.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124341 AK011517.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151860 - -
          Whole Mouse Genome G4122A A_52_P148301 BC024838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148301 BC100548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148301 AK054348.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148301 AK154180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P148301 NM_153121.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P136720 BC024838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P136720 BC100548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P136720 BC138476.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P136720 BC145725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P136720 AK054348.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173233 BC024838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173233 BC100548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173233 AK154180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173233 NM_153121.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173233 AK011517.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22342371 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10401941 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494173 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401940 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401943 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494172 - -
          Affymetrix Mouse Gene 1.0 ST Array 10401942 - -
          Affymetrix HT MG-430 PM Array Plate 1460438_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10401939 - -
          Affymetrix Mouse Gene 1.1 ST Array 10494170 - -
          Mouse Genome 430 2.0 Array 1460438_at - -
          Mouse Genome 430 2.0 Array 1460438_PM_at - -
          Mouse Expression Set 430 1460438_PM_at - -
          Mouse Expression Set 430 1460438_at - -
          Murine Genome U74v2 Set 93165_PM_at - -
          Murine Genome U74v2 Set 93165_at - -
          Mouse Genome 430A 2.0 Array 1460438_PM_at - -
          Mouse Genome 430A 2.0 Array 1460438_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 351482 - -
          ABI Mouse Genome Survey Microarray 737110 - -
          ABI Mouse Genome Survey Microarray 742165 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6350524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238939 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238939 - -
          mouse-6 v1.1 (Illumina) scl0217779.3_307-S - -
          Illumina Sentrix mouse-6.v1 bead array 6350524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0217779.3_307-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238939 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3142193 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3142193 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238939 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0217779.3_307-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline