Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00783690_s1
          See other ADAM26A GE Assays ›
          Gene Symbol:
          Adam26a
          Gene Name:
          a disintegrin and metallopeptidase domain 26A (testase 3)
          Gene Aliases:
          Adam26, Dtgn4
          Chromosome Location:
          Chr.8: 43568273 - 43576707 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00783690_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adam26a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010085.2 NP_034215.2 2 2429 146
          GenBank mRNA AF167404.1 - 2 2388 146
          AK029756.1 - 2 2430 146

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adam26a

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          multicellular organism development
          spermatogenesis
          cell differentiation
          metalloendopeptidase activity
          integrin binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373065 AK015931.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373065 NM_001009547.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373065 AK015931.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P373065 NM_001009547.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P152463 AF167404.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P152463 AK029756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P152463 NM_010085.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P152463 AF167404.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P152463 AK029756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P152463 NM_010085.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397181 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 221380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99808 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126343 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373065 AK015931.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373065 NM_001009547.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373065 AK015931.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P373065 NM_001009547.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P152463 AF167404.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P152463 AK029756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P152463 NM_010085.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P152463 AF167404.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P152463 AK029756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P152463 NM_010085.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P152463 AF167404.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P152463 AK029756.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P152463 NM_010085.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P152463 AF167404.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P152463 AK029756.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P152463 NM_010085.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25820854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25820862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25820866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25820863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25820865 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126343 - -
          Whole Mouse Genome G4122A A_52_P373065 AK015931.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373065 NM_001009547.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373065 AK015931.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P373065 NM_001009547.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152463 AF167404.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152463 AK029756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152463 NM_010085.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152463 AF167404.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152463 AK029756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P152463 NM_010085.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365571 BC141203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365571 BC141204.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365571 BC132102.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365571 BC132292.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365571 AF373288.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126343 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07006470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07006459 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07006461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07006464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820862 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10578422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578412 - -
          Affymetrix HT MG-430 PM Array Plate 1425395_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10578410 - -
          Affymetrix Mouse Gene 1.1 ST Array 10578421 - -
          Mouse Genome 430 2.0 Array 1425395_at - -
          Mouse Genome 430 2.0 Array 1425395_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13525_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13525_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800016406.mm.1 NM_010085.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800016406.mm.1 TC0800002132.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800016405.mm.1 NM_010085.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800016405.mm.1 TC0800002132.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800016404.mm.1 NM_010085.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800016404.mm.1 TC0800002132.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800016407.mm.1 NM_010085.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800016407.mm.1 TC0800002132.mm.1 MTA TC
          Mouse Expression Set 430 1425395_PM_at - -
          Mouse Expression Set 430 1425395_at - -
          Mouse Genome 430A 2.0 Array 1425395_at - -
          Mouse Genome 430A 2.0 Array 1425395_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425395_at - -
          Affymetrix HT Mouse Genome 430A Array 1425395_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 398007 - -
          ABI Mouse Genome Survey Microarray 401959 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1340047 - -
          Illumina mouse-6 v1.0 expression beadchip 1340047 - -
          Illumina mouse-6 v1.0 expression beadchip 6180736 - -
          Illumina mouse-6 v1.0 expression beadchip 1780026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229860 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244723 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229860 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244723 - -
          mouse-6 v1.1 (Illumina) scl34875.2.1_170-S - -
          mouse-6 v1.1 (Illumina) scl013525.1_8-S - -
          mouse-6 v1.1 (Illumina) scl0013525.1_185-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340047 - -
          Illumina Sentrix mouse-6.v1 bead array 6180736 - -
          Illumina Sentrix mouse-6.v1 bead array 1780026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013525.1_8-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0013525.1_185-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34875.2.1_170-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780026 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180736 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244723 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3004858 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229860 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2622239 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229860 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244723 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3004858 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline