Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00783747_sH
          See other KLRA12 GE Assays ›
          Gene Symbol:
          Klra12
          Gene Name:
          killer cell lectin-like receptor subfamily A, member 12
          Gene Aliases:
          Ly49L, Ly49l1, Ly49l2, Ly49l3, Ly49l4, Ly49p/d
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00783747_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010646.1 NP_034776.1 5 651 125
          GenBank mRNA AF204265.1 - 6 642 125
          AF204266.1 - 5 603 125
          AF204267.1 - 6 633 125
          AF204268.1 - 5 594 125
          AF307947.1 - 5 655 125
          AF307948.1 - 5 651 125

          Back To Top

          More Information


          Updated Mapping Information

          Based on assay re-evaluation in August 2010 with NCBI transcript and genome annotations (Genome Build NCBIM37.1), this assay is now annotated to be across exon-exon junction.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra12

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2167714 AF204265.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2167714 AF204266.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2167714 AF204267.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2167714 NM_010646.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2167714 AF204268.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174017 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112911 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122043 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2167714 AF204265.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2167714 AF204266.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2167714 AF204267.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2167714 NM_010646.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2167714 AF204268.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24847518 - -
          Whole Mouse Genome G4122A A_52_P288892 AF204265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288892 AF204266.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288892 AF204267.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288892 NM_010646.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P288892 AF204268.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131844 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100029 U10093.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100029 U10095.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100029 NM_001110323.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133120 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129479 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139537 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115542 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05164899 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05164897 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05132004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24846359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05229706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05132004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24847518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846786 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548482 - -
          Affymetrix HT MG-430 PM Array Plate 1425453_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1425259_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10548455 - -
          Mouse Genome 430 2.0 Array 1451664_PM_x_at - -
          Mouse Genome 430 2.0 Array 1425259_PM_x_at - -
          Mouse Genome 430 2.0 Array 1425259_x_at - -
          Mouse Genome 430 2.0 Array 1425453_x_at - -
          Mouse Genome 430 2.0 Array 1425453_PM_x_at - -
          Mouse Genome 430 2.0 Array 1451664_x_at - -
          Mouse Expression Set 430 1425259_PM_x_at - -
          Mouse Expression Set 430 1425259_x_at - -
          Mouse Expression Set 430 1425453_PM_x_at - -
          Mouse Expression Set 430 1425453_x_at - -
          Murine Genome U74v2 Set 100387_PM_f_at - -
          Murine Genome U74v2 Set 100387_f_at - -
          Mouse Genome 430A 2.0 Array 1425453_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1425259_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1425259_x_at - -
          Mouse Genome 430A 2.0 Array 1425453_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425259_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425453_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425259_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425453_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 360026 - -
          Illumina mouse-6 v1.0 expression beadchip 1240538 - -
          Illumina mouse-6 v1.0 expression beadchip 100840278 - -
          Illumina mouse-6 v1.0 expression beadchip 360026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001240538 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254500 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254500 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254647 - -
          mouse-6 v1.1 (Illumina) scl0016630.1_215-S - -
          mouse-6 v1.1 (Illumina) GI_21327664-S - -
          mouse-6 v1.1 (Illumina) scl016630.5_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 1240538 - -
          Illumina Sentrix mouse-6.v1 bead array 100840278 - -
          Illumina Sentrix mouse-6.v1 bead array 360026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0016630.1_215-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl016630.5_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360026 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100840278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240538 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2771043 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254500 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2911776 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254500 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2771043 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2911776 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254647 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline