Search
Search
| Gene Symbol: Rab7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293652.1 | NP_001280581.1 | 8 | 1726 | 108 | |
| NM_001293653.1 | NP_001280582.1 | 6 | 1387 | 108 | ||
| NM_001293654.1 | NP_001280583.1 | 7 | 1553 | 108 | ||
| NM_001293655.1 | NP_001280584.1 | 6 | 1466 | 108 | ||
| NM_009005.3 | NP_033031.2 | 6 | 1504 | 108 | ||
| GenBank mRNA | AK005004.1 | - | 6 | 1467 | 108 | |
| AK057671.1 | - | 6 | 1591 | 108 | ||
| AK135659.1 | - | 7 | 1556 | 108 | ||
| AK148117.1 | - | 6 | 1498 | 108 | ||
| AK154234.1 | - | 6 | 1485 | 108 | ||
| AK158971.1 | - | 6 | 1388 | 108 | ||
| AK159507.1 | - | 6 | 1433 | 108 | ||
| AK159928.1 | - | 6 | 1405 | 108 | ||
| BC004597.1 | - | 6 | 1422 | 3584891 | 108 | |
| X89650.1 | - | 6 | 1450 | 108 | ||
Set Membership: |
|