Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00784451_s1
          See other COX7A2L GE Assays ›
          Gene Symbol:
          Cox7a2l
          Gene Name:
          cytochrome c oxidase subunit VIIa polypeptide 2-like
          Gene Aliases:
          COX7AR, COX7RP, EB1, SIG-81, SIG81, Silg81
          Chromosome Location:
          Chr.17: 83501917 - 83514333 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00784451_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox7a2l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159529.1 NP_001153001.1 4 854 146
          NM_009187.3 NP_033213.1 3 785 146
          GenBank mRNA AK009614.1 - 4 851 145
          AK133520.1 - 3 785 146
          AK150034.1 - 3 782 146
          AK152553.1 - 3 775 146
          AK159441.1 - 3 783 146
          AK166761.1 - 3 770 146
          AK167062.1 - 3 780 146
          AK169619.1 - 3 804 145
          BC004715.1 - 3 756 3582142 146
          BC085296.1 - 3 761 5337317 146
          BP769290.1 - 1 146
          X80899.1 - 3 752 146

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Mouse Estrogen Panel 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox7a2l

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of oxidative phosphorylation
          mitochondrial respiratory chain supercomplex assembly
          cytochrome-c oxidase activity
          electron carrier activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399350 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399354 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399355 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399351 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399357 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399352 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129419 X80899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151071 X80899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151071 AK169619.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151071 BC004715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151071 BC085296.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151071 BP769290.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10477 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35113 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162048 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150498 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151071 X80899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151071 AK169619.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151071 BC004715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151071 BC085296.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151071 BP769290.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129419 X80899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102569 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434803 X80899.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434803 AK169619.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434803 BC004715.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434803 BC085296.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434803 AK133520.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399377 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31399361 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03383213 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150498 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162048 - -
          Whole Mouse Genome G4122A A_51_P434803 X80899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434803 AK169619.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434803 BC004715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434803 BC085296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434803 AK133520.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116016 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162048 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150498 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568060 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568070 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568066 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13568067 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399355 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399350 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399354 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399351 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399352 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31399357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31399344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10410283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28290748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28290743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10410279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28290747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13317054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13435626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13554659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13323336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13227953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13379136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13379135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13307229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13425959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13119279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13577376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13568060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13234667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13307242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13568064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13425957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31399391 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10453255 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399679 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399678 - -
          Affymetrix HT MG-430 PM Array Plate 1432263_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1432264_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1421772_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10399677 - -
          Affymetrix Mouse Gene 1.1 ST Array 10453254 - -
          Mouse Genome 430 2.0 Array 1432264_x_at - -
          Mouse Genome 430 2.0 Array 1421772_PM_a_at - -
          Mouse Genome 430 2.0 Array 1432263_PM_a_at - -
          Mouse Genome 430 2.0 Array 1421772_a_at - -
          Mouse Genome 430 2.0 Array 1432263_a_at - -
          Mouse Genome 430 2.0 Array 1432264_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20463_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20463_at - -
          Mouse Expression Set 430 1421772_a_at - -
          Mouse Expression Set 430 1432264_x_at - -
          Mouse Expression Set 430 1421772_PM_a_at - -
          Mouse Expression Set 430 1432264_PM_x_at - -
          Mouse Expression Set 430 1432263_a_at - -
          Mouse Expression Set 430 1432263_PM_a_at - -
          Murine Genome U74v2 Set 160383_at - -
          Murine Genome U74v2 Set 160383_PM_at - -
          Mouse Genome 430A 2.0 Array 1432263_a_at - -
          Mouse Genome 430A 2.0 Array 1432263_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1432264_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1421772_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1432264_x_at - -
          Mouse Genome 430A 2.0 Array 1421772_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432264_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1432264_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1432263_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421772_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421772_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1432263_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 533479 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1850452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2888713 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2888713 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline