Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00785614_sH
          See other VPREB1 GE Assays ›
          Gene Symbol:
          Vpreb1
          Gene Name:
          pre-B lymphocyte gene 1
          Gene Aliases:
          CD179a, Vpreb-1
          Chromosome Location:
          Chr.16: 16868401 - 16869255 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00785614_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vpreb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016982.2 NP_058678.1 2 459 128
          GenBank mRNA AK010843.1 - 2 453 128
          AK010879.1 - 2 453 128
          BC117051.1 - 2 421 40126370 128
          BC119175.1 - 1 508 8733924 128
          BC144850.1 - 1 508 9053398 128
          BE852604.1 - 1 128
          X05556.1 - 2 454 128

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vpreb1

          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          immunoglobulin production
          immune response
          regulation of cell size
          hemopoiesis
          B cell proliferation
          homeostasis of number of cells
          receptor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30570743 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30570744 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 BC117051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 BC119175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 BC144850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 BE852604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 X05556.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 BC117051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 BC119175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 BC144850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 BE852604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P400217 X05556.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 BC117051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 BC119175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 BC144850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 BE852604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 X05556.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 BC117051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 BC119175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 BC144850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 BE852604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P400217 X05556.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 BC117051.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 BC119175.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 BC144850.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 BE852604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 X05556.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 BC117051.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 BC119175.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 BC144850.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 BE852604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P400217 X05556.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30570740 - -
          Whole Mouse Genome G4122A A_51_P400217 BC117051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400217 BC119175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400217 BC144850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400217 BE852604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400217 X05556.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400217 BC117051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400217 BC119175.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400217 BC144850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400217 BE852604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P400217 X05556.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119830 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127805 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12697673 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12802900 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12697669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12802894 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12802906 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12943266 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30570743 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30570744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30570772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12583880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12583894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12802894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12569149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12914252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12788755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12802900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12583876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12583879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12583900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12856712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12587630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30570707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12587631 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438067 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438066 - -
          Affymetrix HT MG-430 PM Array Plate 1449869_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10438064 - -
          Mouse Genome 430 2.0 Array 1422416_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449869_at - -
          Mouse Genome 430 2.0 Array 1449869_PM_at - -
          Mouse Genome 430 2.0 Array 1422416_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22362_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22362_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600010293.mm.1 TC1600001271.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010291.mm.1 NM_016982.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010291.mm.1 NM_016982.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010291.mm.1 TC1600001271.mm.1 MTA TC
          Mouse Expression Set 430 1449869_at - -
          Mouse Expression Set 430 1422416_s_at - -
          Mouse Expression Set 430 1422416_PM_s_at - -
          Mouse Expression Set 430 1449869_PM_at - -
          Murine Genome U74v2 Set 161297_f_at - -
          Murine Genome U74v2 Set 161297_PM_f_at - -
          Murine Genome U74v2 Set 92972_PM_at - -
          Murine Genome U74v2 Set 92972_at - -
          Mouse Genome 430A 2.0 Array 1422416_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1449869_PM_at - -
          Mouse Genome 430A 2.0 Array 1422416_s_at - -
          Mouse Genome 430A 2.0 Array 1449869_at - -
          Affymetrix HT Mouse Genome 430A Array 1449869_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449869_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 908099 - -
          ABI Mouse Genome Survey Microarray 917607 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3520280 - -
          Illumina mouse-6 v1.0 expression beadchip 3520280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2489226 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2489226 - -
          mouse-6 v1.1 (Illumina) scl0022362.1_271-S - -
          Illumina Sentrix mouse-6.v1 bead array 3520280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0022362.1_271-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2846485 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2846485 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0022362.1_271-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline