Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00786031_s1
          See other RPL7L1 GE Assays ›
          Gene Symbol:
          Rpl7l1
          Gene Name:
          ribosomal protein L7-like 1
          Gene Aliases:
          1500016H10Rik
          Chromosome Location:
          Chr.17: 46773907 - 46782656 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00786031_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl7l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025433.3 NP_079709.2 7 2365 134
          GenBank mRNA AK090082.1 - 7 2365 134
          AK152288.1 - 6 2294 134
          AK167036.1 - 7 2365 134

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl7l1

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst formation
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 BC003907.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 BC030165.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 AK152288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 AK166499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 AK167036.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 BC003907.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 BC030165.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 AK152288.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 AK166499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P343343 AK167036.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287758 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 AK134287.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 AK146442.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 AK152420.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 AK152931.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 AK159933.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 AK159950.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 AK163373.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 AK191486.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 BC004794.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431551 NM_145583.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 372457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77119 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152038 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 BC003907.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 BC030165.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 AK152288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 AK166499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 AK167036.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 BC003907.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 BC030165.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 AK152288.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 AK166499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P343343 AK167036.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110147 BC003907.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110147 AK152288.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110147 AK167036.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110147 AK090082.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P110147 NM_025433.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31208905 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152038 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08790795 - -
          Whole Mouse Genome G4122A A_51_P110147 BC003907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110147 AK152288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110147 AK167036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110147 AK090082.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P110147 NM_025433.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 BC003907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 BC030165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 AK152288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 AK166499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 AK167036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 BC003907.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 BC030165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 AK152288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 AK166499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343343 AK167036.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135519 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103451 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152038 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08790795 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13240102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13120414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13120416 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13240097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31208862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13259198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13412155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13418250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13332721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13412160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13290297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13474374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31208907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13119942 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10451475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10451479 - -
          Affymetrix HT MG-430 PM Array Plate 1426386_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452049_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439780_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10451472 - -
          Mouse Genome 430 2.0 Array 1439780_at - -
          Mouse Genome 430 2.0 Array 1452049_at - -
          Mouse Genome 430 2.0 Array 1439780_PM_at - -
          Mouse Genome 430 2.0 Array 1426386_PM_at - -
          Mouse Genome 430 2.0 Array 1426386_at - -
          Mouse Genome 430 2.0 Array 1452049_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66229_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66229_at - -
          Mouse Expression Set 430 1426386_at - -
          Mouse Expression Set 430 1452049_at - -
          Mouse Expression Set 430 1426386_PM_at - -
          Mouse Expression Set 430 1452049_PM_at - -
          Mouse Expression Set 430 1439780_PM_at - -
          Mouse Expression Set 430 1439780_at - -
          Murine Genome U74v2 Set 98138_PM_at - -
          Murine Genome U74v2 Set 98138_at - -
          Mouse Genome 430A 2.0 Array 1452049_at - -
          Mouse Genome 430A 2.0 Array 1426386_at - -
          Mouse Genome 430A 2.0 Array 1426386_PM_at - -
          Mouse Genome 430A 2.0 Array 1452049_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452049_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452049_at - -
          Affymetrix HT Mouse Genome 430A Array 1426386_at - -
          Affymetrix HT Mouse Genome 430A Array 1426386_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439780_at - -
          Affymetrix HT Mouse Genome 430B Array 1439780_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 905135 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3870075 - -
          MouseWG-6 v2 Expression BeadChip 610010 - -
          Illumina mouse-6 v1.0 expression beadchip 103360092 - -
          Illumina mouse-6 v1.0 expression beadchip 3870075 - -
          Illumina mouse-6 v1.0 expression beadchip 610010 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610010 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2661407 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249606 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252768 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2661407 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249606 - -
          mouse-6 v1.1 (Illumina) GI_27754133-S - -
          mouse-6 v1.1 (Illumina) scl066229.1_118-S - -
          mouse-6 v1.1 (Illumina) scl0066229.1_167-S - -
          Illumina Sentrix mouse-6.v1 bead array 103360092 - -
          Illumina Sentrix mouse-6.v1 bead array 3870075 - -
          Illumina Sentrix mouse-6.v1 bead array 610010 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066229.1_118-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066229.1_167-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103360092 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610010 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249606 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2661407 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2661407 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249606 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0066229.1_167-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066229.1_118-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0