Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00787005_s1
          See other HIGD1A GE Assays ›
          Gene Symbol:
          Higd1a
          Gene Name:
          HIG1 domain family, member 1A
          Gene Aliases:
          2210020B17Rik, 7420700H20Rik, AI303276, AW049839, HIMP1, Hig1
          Chromosome Location:
          Chr.9: 121848560 - 121858357 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00787005_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Higd1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019814.4 NP_062788.1 4 1809 117
          GenBank mRNA AK008756.1 - 4 1436 117
          AY028386.1 - 4 1363 117
          AY028387.1 - 3 1869 117

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Higd1a

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          cellular response to glucose starvation
          negative regulation of apoptotic process
          oxidation-reduction process
          cellular response to hypoxia
          negative regulation of release of cytochrome c from mitochondria
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886026 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886025 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886027 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886031 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886028 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 BC132371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 BC151191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 BE457620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 AK018360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 AY028386.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 BC132371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 BC151191.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 BE457620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 AK018360.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P458697 AY028386.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926197 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731385 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89682 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153177 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 BC132371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 BC151191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 BE457620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 AK018360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 AY028386.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 BC132371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 BC151191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 BE457620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 AK018360.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P458697 AY028386.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263310 AB253941.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P263310 AK008756.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886021 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153177 - -
          Whole Mouse Genome G4122A A_51_P263310 AB253941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P263310 AK008756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 BC132371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 BC151191.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 BE457620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 AK018360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 AY028386.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 BC132371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 BC151191.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 BE457620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 AK018360.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458697 AY028386.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122470 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106712 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153177 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07600759 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07600761 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07600754 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07600765 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886025 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886026 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886031 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886029 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26886027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07981872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08018996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07600765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08051858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886040 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10597874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10492863 - -
          Affymetrix HT MG-430 PM Array Plate 1448359_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1416481_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10492860 - -
          Affymetrix Mouse Gene 1.1 ST Array 10597871 - -
          Mouse Genome 430 2.0 Array 1416481_PM_s_at - -
          Mouse Genome 430 2.0 Array 1416480_a_at - -
          Mouse Genome 430 2.0 Array 1416480_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448359_a_at - -
          Mouse Genome 430 2.0 Array 1416481_s_at - -
          Mouse Genome 430 2.0 Array 1448359_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56295_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56295_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900027634.mm.1 NM_019814.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027634.mm.1 NM_019814.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027634.mm.1 TC0900003275.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027635.mm.1 NM_019814.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027635.mm.1 NM_019814.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027635.mm.1 TC0900003275.mm.1 MTA TC
          Mouse Expression Set 430 1416481_PM_s_at - -
          Mouse Expression Set 430 1416480_PM_a_at - -
          Mouse Expression Set 430 1448359_PM_a_at - -
          Mouse Expression Set 430 1416480_a_at - -
          Mouse Expression Set 430 1416481_s_at - -
          Mouse Expression Set 430 1448359_a_at - -
          Murine Genome U74v2 Set 161976_PM_r_at - -
          Murine Genome U74v2 Set 101094_at - -
          Murine Genome U74v2 Set 101094_PM_at - -
          Murine Genome U74v2 Set 161976_r_at - -
          Mouse Genome 430A 2.0 Array 1416481_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1448359_a_at - -
          Mouse Genome 430A 2.0 Array 1416480_a_at - -
          Mouse Genome 430A 2.0 Array 1416480_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416481_s_at - -
          Mouse Genome 430A 2.0 Array 1448359_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448359_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416481_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1416481_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1448359_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 502257 - -
          ABI Mouse Genome Survey Microarray 504962 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1190239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2644630 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2644630 - -
          mouse-6 v1.1 (Illumina) scl35194.5_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190239 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35194.5_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2644630 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2644630 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35194.5_5-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0