Search
Search

Col1a1| Gene Symbol: Col1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007742.3 | NP_031768.2 | 3 - 4 | 403 | 123 | |
| GenBank mRNA | AK132180.1 | - | 3 - 4 | 419 | 123 | |
| AK158011.1 | - | 3 - 4 | 419 | 123 | ||
| AK159283.1 | - | 3 - 4 | 421 | 123 | ||
| AK159285.1 | - | 3 - 4 | 423 | 123 | ||
| BC050014.1 | - | 3 - 4 | 403 | 4225038 | 123 | |
| BC059281.1 | - | 3 - 4 | 413 | 3992363 | 123 | |
| U08020.1 | - | 3 - 4 | 304 | 123 | ||
Set Membership: |
|