| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500683 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500679 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500678 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500682 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500684 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500729 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500727 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500730 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500680 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500728 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2104219 |
AF193796.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2104219 |
NM_010464.2 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
332052 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
437821 |
AK033743.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
437821 |
NM_177840.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
437821 |
XM_885099.2 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
437821 |
XM_913732.2 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
151032 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
939738 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
58400 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P166302 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2104219 |
AF193796.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2104219 |
NM_010464.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P378277 |
AF193796.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500660 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500646 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500633 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500668 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500661 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500693 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500686 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500648 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500632 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500654 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500680 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500649 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500676 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500640 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500690 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500643 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500638 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500687 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500695 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500657 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500641 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500673 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500670 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500677 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500664 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500651 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500666 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500672 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500691 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500655 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500662 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500636 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500683 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500659 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500645 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500663 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500692 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500652 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500684 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500667 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500669 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500653 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30500681 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P166302 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03119338 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P378277 |
AF193796.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124846 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P166302 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12400452 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12317191 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12400456 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12400454 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12317201 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12108539 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500678 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500727 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500684 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500729 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500682 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500683 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500730 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500728 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500680 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30500679 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500603 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500683 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500811 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500620 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500660 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500774 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500780 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500813 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500778 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500809 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500723 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500798 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500739 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500821 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500627 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500771 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500648 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500677 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500629 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500649 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500640 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500645 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500792 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500616 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500676 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500622 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500722 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500759 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500807 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500609 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500651 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500653 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500802 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500786 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500818 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500732 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500662 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500740 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500664 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500663 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500750 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500654 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500763 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500766 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500815 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500619 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500715 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500701 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500607 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500612 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500734 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500816 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500697 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500636 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500625 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500623 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500643 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500765 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500735 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500641 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500717 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500705 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500704 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500710 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500761 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500684 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500754 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500608 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500805 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500822 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500784 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500801 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500825 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500618 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500707 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500614 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500667 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500598 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500670 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500605 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500751 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500787 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500752 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500638 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500693 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500672 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500673 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500728 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500726 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500799 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500713 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500756 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500711 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500666 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500820 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500633 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500680 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500824 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500703 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500719 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500659 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500699 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500686 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500768 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500737 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500657 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500730 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500777 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500695 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500602 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500681 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500691 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500668 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500687 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500770 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500790 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500797 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500794 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30500646 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500693 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500683 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500735 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500599 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12303629 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500699 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500651 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500672 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500695 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500619 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500715 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500657 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500661 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500645 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500721 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500703 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500684 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500659 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500722 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500664 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500616 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500690 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500739 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500623 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500680 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11923959 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500705 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12459416 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500701 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12303620 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12303772 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500600 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500713 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500737 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500660 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500627 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500710 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500732 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500670 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12340573 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500649 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12303625 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500673 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500740 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500640 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12317201 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500638 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500666 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12317197 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500676 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500618 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12302596 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500687 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500632 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500633 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500686 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500719 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500704 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500667 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500629 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500707 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12302604 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500734 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500681 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500630 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12302603 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500622 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500614 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500636 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500717 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500723 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500625 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500697 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500620 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30500598 |
- |
- |