Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00802869_g1
          See other IRS3 GE Assays ›
          Gene Symbol:
          Irs3
          Gene Name:
          insulin receptor substrate 3
          Gene Aliases:
          AA408304
          Chromosome Location:
          Chr.5: 137643030 - 137645748 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00802869_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Irs3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010571.3 NP_034701.1 1 - 2 629 65
          GenBank mRNA AF035948.1 - 1 - 2 435 65
          AK030311.1 - 1 - 2 631 65
          AK040947.1 - 1 - 2 614 65
          BC098235.1 - 1 - 2 461 1546952 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003451.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Irs3

          Gene Ontology Categories:

          Function(s) Process(es)

          insulin receptor signaling pathway
          regulation of lipid metabolic process
          signal transducer activity
          insulin receptor binding
          phosphatidylinositol 3-kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126843 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126845 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2214841 BC098235.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2214841 NM_010571.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2214841 AK030311.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174972 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2214841 BC098235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2214841 NM_010571.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2214841 AK030311.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174158 AK040947.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174158 NM_010571.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174158 AF035948.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174158 AK030311.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24126871 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05761166 - -
          Whole Mouse Genome G4122A A_51_P174158 AK040947.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174158 NM_010571.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174158 AF035948.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174158 AK030311.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133609 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05761166 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04820354 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04820360 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04820353 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04820363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04820352 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04820356 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04820358 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126843 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126845 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24126846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24126811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04210524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04820360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04210519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04820359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04763347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04820353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04250655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04250658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04632247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04820356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04210523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04469150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04820363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04361479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04632250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04210525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24126841 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534886 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534888 - -
          Affymetrix HT MG-430 PM Array Plate 1437672_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534883 - -
          Mouse Genome 430 2.0 Array 1437672_PM_at - -
          Mouse Genome 430 2.0 Array 1437672_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16369_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16369_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500031481.mm.1 TC0500003451.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031481.mm.1 NM_010571.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031481.mm.1 NM_010571.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031483.mm.1 TC0500003451.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031483.mm.1 NM_010571.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031483.mm.1 NM_010571.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031482.mm.1 TC0500003451.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031482.mm.1 NM_010571.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031482.mm.1 NM_010571.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031484.mm.1 TC0500003451.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031484.mm.1 NM_010571.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031484.mm.1 NM_010571.3 Refseq
          Mouse Expression Set 430 1437672_PM_at - -
          Mouse Expression Set 430 1437672_at - -
          Mouse Genome 430A 2.0 Array 1437672_at - -
          Mouse Genome 430A 2.0 Array 1437672_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1437672_at - -
          Affymetrix HT Mouse Genome 430A Array 1437672_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 704213 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6550452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755312 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2755312 - -
          mouse-6 v1.1 (Illumina) scl25874.2_338-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25874.2_338-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2755312 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755312 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0