Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00802925_m1
          See other TMED1 GE Assays ›
          Gene Symbol:
          Tmed1
          Gene Name:
          transmembrane emp24 domain containing 1
          Gene Aliases:
          Il1rl1l, Ly84l, St2l
          Chromosome Location:
          Chr.9: 21507380 - 21510187 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00802925_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmed1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010744.3 NP_034874.2 1 - 2 193 79
          GenBank mRNA AK133513.1 - 1 - 2 193 79
          BC016568.1 - 1 - 2 194 79
          BC138498.1 - 1 - 2 241 8861518 79
          BC138499.1 - 1 - 2 240 8861519 79
          BC145298.1 - 1 - 2 241 79
          U41805.1 - 1 - 2 169 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001874.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmed1

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s)

          transport
          protein transport

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347558 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347560 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347559 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983558 BC016568.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983558 BC006715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983558 NM_010744.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983558 AK133513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983558 AK160525.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920686 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115594 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983558 BC016568.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983558 BC006715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983558 NM_010744.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983558 AK133513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983558 AK160525.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P221852 BC016568.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P221852 BC006715.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P221852 NM_010744.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P221852 AK133513.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P221852 AK160525.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26347563 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115594 - -
          Whole Mouse Genome G4122A A_51_P221852 BC016568.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221852 BC006715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221852 NM_010744.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221852 AK133513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P221852 AK160525.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113553 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115594 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08162344 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08162350 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08162352 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08162351 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08162346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347556 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347560 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347558 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26347559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26347552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07929652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07929622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08162350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07929639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07751920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08162346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26347565 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591539 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591541 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591538 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591543 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591540 - -
          Affymetrix HT MG-430 PM Array Plate 1451327_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10583647 - -
          Affymetrix Mouse Gene 1.1 ST Array 10591537 - -
          Mouse Genome 430 2.0 Array 1451327_PM_a_at - -
          Mouse Genome 430 2.0 Array 1451327_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17083_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17083_at - -
          Affymetrix Mouse Clariom S Array PSR0900015764.mm.1 NM_010744.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015764.mm.1 TC0900001874.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900015767.mm.1 TC0900001874.mm.1 MTA TC
          Mouse Expression Set 430 1451327_PM_a_at - -
          Mouse Expression Set 430 1451327_a_at - -
          Murine Genome U74v2 Set 162484_r_at - -
          Murine Genome U74v2 Set 162484_PM_r_at - -
          Murine Genome U74v2 Set 160061_at - -
          Murine Genome U74v2 Set 160061_PM_at - -
          Mouse Genome 430A 2.0 Array 1451327_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1451327_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451327_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451327_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 931758 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5910717 - -
          Illumina mouse-6 v1.0 expression beadchip 3140685 - -
          Illumina mouse-6 v1.0 expression beadchip 5910717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003140685 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249428 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2738318 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249428 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2738318 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224114 - -
          mouse-6 v1.1 (Illumina) scl36139.4_125-S - -
          mouse-6 v1.1 (Illumina) scl0003607.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 5910717 - -
          Illumina Sentrix mouse-6.v1 bead array 3140685 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36139.4_125-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003607.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3140685 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249428 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1214974 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249428 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214974 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline