Search
Search
| Gene Symbol: Myl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010859.2 | NP_034989.1 | 3 - 4 | 397 | 139 | |
| GenBank mRNA | AK002312.1 | - | 3 - 4 | 399 | 139 | |
| AK011518.1 | - | 3 - 4 | 390 | 139 | ||
| AK146943.1 | - | 3 - 4 | 401 | 139 | ||
| BC061222.1 | - | 3 - 4 | 400 | 30297023 | 139 | |
Set Membership: |
|