Search
Search

Ppard| Gene Symbol: Ppard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011145.3 | NP_035275.1 | 5 - 6 | 696 | 68 | |
| GenBank mRNA | AB221579.1 | - | 3 - 4 | 425 | 68 | |
| AK089913.1 | - | 5 - 6 | 644 | 68 | ||
| BC070398.1 | - | 5 - 6 | 647 | 5694282 | 68 | |
| L28116.1 | - | 3 - 4 | 478 | 68 | ||
| U10375.1 | - | 3 - 4 | 421 | 68 | ||
Set Membership: |
|