Search
Search

Scnn1a| Gene Symbol: Scnn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011324.2 | NP_035454.2 | 2 - 3 | 1310 | 69 | |
| GenBank mRNA | AF112185.1 | - | 2 - 3 | 797 | 69 | |
| AK140389.1 | - | 2 - 3 | 1311 | 69 | ||
| BC040814.1 | - | 2 - 3 | 940 | 69 | ||
| BC133688.1 | - | 2 - 3 | 791 | 40111601 | 69 | |
| BC133689.1 | - | 2 - 3 | 791 | 40111602 | 69 | |
Set Membership: |
|