Search
Search

Abcc8| Gene Symbol: Abcc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011510.3 | NP_035640.2 | 28 - 29 | 3668 | 77 | |
| GenBank mRNA | AK083916.1 | - | 19 - 20 | 2083 | 77 | |
| AK085812.1 | - | 3 - 4 | 329 | 77 | ||
| BC057080.1 | - | 15 - 16 | 1548 | 77 | ||
| BC141411.1 | - | 28 - 29 | 3652 | 9056024 | 77 | |
Set Membership: |
|