Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00803538_m1
          See other TGFBR3 GE Assays ›
          Gene Symbol:
          Tgfbr3
          Gene Name:
          transforming growth factor, beta receptor III
          Gene Aliases:
          1110036H20Rik, AU015626, AW215636, TBRIII
          Chromosome Location:
          Chr.5: 107106570 - 107289595 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00803538_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tgfbr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011578.3 NP_035708.2 13 - 14 2595 96
          GenBank mRNA AF039601.1 - 13 - 14 96
          AK133234.1 - 11 - 12 2105 96
          BC070428.1 - 13 - 14 2520 6410554 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002919.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tgfbr3

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          vasculogenesis
          osteoblast differentiation
          in utero embryonic development
          blastocyst development
          epithelial to mesenchymal transition
          liver development
          blood vessel remodeling
          epicardial cell to mesenchymal cell transition
          protein complex assembly
          protein phosphorylation
          immune response
          transforming growth factor beta receptor signaling pathway
          transforming growth factor beta receptor complex assembly
          positive regulation of cell proliferation
          embryo development
          negative regulation of epithelial cell migration
          negative regulation of epithelial to mesenchymal transition
          cell growth
          BMP signaling pathway
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of BMP signaling pathway
          response to follicle-stimulating hormone
          response to prostaglandin E
          response to luteinizing hormone
          intracellular signal transduction
          regulation of protein binding
          negative regulation of epithelial cell proliferation
          regulation of blood vessel size
          positive regulation of NF-kappaB transcription factor activity
          negative regulation of cellular component movement
          ventricular cardiac muscle tissue morphogenesis
          palate development
          cardiac muscle cell proliferation
          definitive hemopoiesis
          cardiac epithelial to mesenchymal transition
          definitive erythrocyte differentiation
          heart trabecula formation
          pathway-restricted SMAD protein phosphorylation
          coronary vasculature development
          coronary vasculature morphogenesis
          vasculogenesis involved in coronary vascular morphogenesis
          visceral serous pericardium development
          negative regulation of apoptotic process involved in morphogenesis
          transforming growth factor beta-activated receptor activity
          type II transforming growth factor beta receptor binding
          transforming growth factor beta receptor binding
          protein binding
          glycosaminoglycan binding
          heparin binding
          coreceptor activity
          fibroblast growth factor binding
          PDZ domain binding
          SMAD binding
          activin binding
          transforming growth factor beta binding
          transforming growth factor beta receptor activity, type III

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500025818.mm.1 TC0500002919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025818.mm.1 NM_011578.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025819.mm.1 TC0500002919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025819.mm.1 NM_011578.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025821.mm.1 TC0500002919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025821.mm.1 NM_011578.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025820.mm.1 TC0500002919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025820.mm.1 NM_011578.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025816.mm.1 TC0500002919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025816.mm.1 NM_011578.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500025815.mm.1 TC0500002919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500025815.mm.1 NM_011578.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline