Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00803754_g1
          See other COPS6 GE Assays ›
          Gene Symbol:
          Cops6
          Gene Name:
          COP9 (constitutive photomorphogenic) homolog, subunit 6 (Arabidopsis thaliana)
          Gene Aliases:
          Sgn3, VIP/MOV34
          Chromosome Location:
          Chr.5: 138161102 - 138163984 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00803754_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cops6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012002.3 NP_036132.1 5 - 6 486 94
          GenBank mRNA AF071315.1 - 5 - 6 523 94
          AK138409.1 - 4 - 5 612 94
          AK146775.1 - 5 - 6 499 94
          BC004664.1 - 5 - 6 485 3498214 94
          BC014286.1 - 5 - 6 490 4163277 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001643.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cops6

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cullin deneddylation
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969042 BC014286.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969042 AK138409.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969042 BC004664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1969042 NM_012002.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128373 BC014286.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128373 AF071315.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128373 AK138409.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128373 AK146410.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128373 AK146775.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807832 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123597 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969042 BC014286.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969042 AK138409.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969042 BC004664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1969042 NM_012002.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128373 BC014286.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128373 AF071315.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128373 AK138409.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128373 AK146410.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2128373 AK146775.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129077 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123597 - -
          Whole Mouse Genome G4122A A_51_P151972 BC014286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151972 AF071315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151972 AK138409.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151972 AK146410.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P151972 AK146775.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P667177 BC014286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P667177 AF071315.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P667177 AK138409.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P667177 AK146775.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P667177 BC004664.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126383 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123597 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04397079 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04397081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04109672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04791766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04146930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04229712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04229735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04397081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04229729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04292238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04684838 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526747 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526749 - -
          Affymetrix HT MG-430 PM Array Plate 1451367_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451366_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526743 - -
          Mouse Genome 430 2.0 Array 1451366_at - -
          Mouse Genome 430 2.0 Array 1451367_at - -
          Mouse Genome 430 2.0 Array 1451366_PM_at - -
          Mouse Genome 430 2.0 Array 1451367_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26893_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26893_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500015045.mm.1 TC0500001643.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015045.mm.1 NM_012002.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015045.mm.1 NM_012002.3 Refseq
          Mouse Expression Set 430 1451366_at - -
          Mouse Expression Set 430 1451367_at - -
          Mouse Expression Set 430 1451367_PM_at - -
          Mouse Expression Set 430 1451366_PM_at - -
          Murine Genome U74v2 Set 99106_at - -
          Murine Genome U74v2 Set 99106_PM_at - -
          Mouse Genome 430A 2.0 Array 1451366_at - -
          Mouse Genome 430A 2.0 Array 1451366_PM_at - -
          Mouse Genome 430A 2.0 Array 1451367_PM_at - -
          Mouse Genome 430A 2.0 Array 1451367_at - -
          Affymetrix HT Mouse Genome 430A Array 1451366_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451367_at - -
          Affymetrix HT Mouse Genome 430A Array 1451366_at - -
          Affymetrix HT Mouse Genome 430A Array 1451367_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 927425 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1850195 - -
          Illumina mouse-6 v1.0 expression beadchip 1850195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850195 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2618793 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2618793 - -
          mouse-6 v1.1 (Illumina) scl026893.9_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 1850195 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl026893.9_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850195 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2618793 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2618793 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl026893.9_4-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline