Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cops6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012002.3 | NP_036132.1 | 7 - 8 | 653 | 78 | |
GenBank mRNA | AF071315.1 | - | 7 - 8 | 690 | 78 | |
AK138409.1 | - | 6 - 7 | 779 | 78 | ||
AK146775.1 | - | 7 - 8 | 666 | 78 | ||
BC004664.1 | - | 7 - 8 | 652 | 3498214 | 78 | |
BC014286.1 | - | 7 - 8 | 657 | 4163277 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |