Search
Search

Tulp1| Gene Symbol: Tulp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021478.1 | NP_067453.1 | 13 - 14 | 1354 | 61 | |
| GenBank mRNA | AF085681.1 | - | 13 - 14 | 1402 | 61 | |
| AF105711.1 | - | 13 - 14 | 1402 | 61 | ||
| AK053880.1 | - | 7 - 8 | 857 | 61 | ||
| AK164621.1 | - | 3 - 4 | 358 | 61 | ||
| BC014820.1 | - | 13 - 14 | 1360 | 61 | ||
| BC038073.1 | - | 1 - 2 | 246 | 61 | ||
Set Membership: |
|