Search
Search
| Gene Symbol: Kin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025280.2 | NP_079556.1 | 2 - 3 | 234 | 78 | |
| GenBank mRNA | AK009429.1 | - | 2 - 3 | 266 | 78 | |
| AK132548.1 | - | 2 - 3 | 664 | 78 | ||
| AK167740.1 | - | 2 - 3 | 238 | 78 | ||
| BC028860.1 | - | 2 - 3 | 234 | 3673830 | 78 | |
| BC058169.1 | - | 2 - 3 | 263 | 78 | ||
| X58472.1 | - | 2 - 3 | 239 | 78 | ||
Set Membership: |
|