| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800008753.mm.1 |
NM_026452.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800008753.mm.1 |
NM_026452.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800008753.mm.1 |
TC0800001123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800008759.mm.1 |
NM_026452.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800008759.mm.1 |
NM_026452.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800008759.mm.1 |
TC0800001123.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800008760.mm.1 |
NM_026452.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800008760.mm.1 |
NM_026452.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800008760.mm.1 |
TC0800001123.mm.1 |
MTA TC |