Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00804888_m1
          See other FUT10 GE Assays ›
          Gene Symbol:
          Fut10
          Gene Name:
          fucosyltransferase 10
          Gene Aliases:
          fuc-TX, fut10A
          Chromosome Location:
          Chr.8: 31187331 - 31261924 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00804888_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fut10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012517.5 NP_001012535.1 2 - 3 464 106
          NM_001286422.1 NP_001273351.1 3 - 4 569 106
          NM_001286424.1 NP_001273353.1 2 - 3 357 106
          NM_001286425.1 NP_001273354.1 2 - 3 464 106
          NM_134161.3 NP_598922.1 2 - 3 464 106
          GenBank mRNA AJ880009.1 - 1 - 2 77 106
          AK083041.1 - 2 - 3 453 106
          AK147136.1 - 3 - 4 525 106
          AK169153.1 - 2 - 3 418 106
          BC030474.1 - 2 - 3 361 5368333 106
          BC062113.1 - 2 - 3 463 6816067 106
          BY721538.1 - 2 - 3 465 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000371.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fut10

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          cerebral cortex radially oriented cell migration
          fucosylation
          neuronal stem cell population maintenance
          alpha-1,6-mannosyltransferase activity
          alpha-1,2-mannosyltransferase activity
          mannosyltransferase activity
          alpha-1,3-mannosyltransferase activity
          alpha-1,3-galactosyltransferase activity
          UDP-glucose:glycoprotein glucosyltransferase activity
          glycolipid mannosyltransferase activity
          oligosaccharyl transferase activity
          dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
          acetylglucosaminyltransferase activity
          acetylgalactosaminyltransferase activity
          galactosyltransferase activity
          fucosyltransferase activity
          O antigen polymerase activity
          lipopolysaccharide-1,6-galactosyltransferase activity
          transferase activity
          transferase activity, transferring glycosyl groups
          cellulose synthase activity
          9-phenanthrol UDP-glucuronosyltransferase activity
          1-phenanthrol glycosyltransferase activity
          9-phenanthrol glycosyltransferase activity
          1,2-dihydroxy-phenanthrene glycosyltransferase activity
          phenanthrol glycosyltransferase activity
          beta-1,4-mannosyltransferase activity
          alpha-1,2-galactosyltransferase activity
          dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
          lipopolysaccharide-1,5-galactosyltransferase activity
          dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
          inositol phosphoceramide synthase activity
          glucosyltransferase activity
          alpha-(1->3)-fucosyltransferase activity
          alpha-(1->6)-fucosyltransferase activity
          indole-3-butyrate beta-glucosyltransferase activity
          salicylic acid glucosyltransferase (ester-forming) activity
          salicylic acid glucosyltransferase (glucoside-forming) activity
          benzoic acid glucosyltransferase activity
          chondroitin hydrolase activity
          dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
          cytokinin 9-beta-glucosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800002842.mm.1 NM_001012517.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002842.mm.1 NM_001286422.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002842.mm.1 NM_001286424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002842.mm.1 NM_001286425.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002842.mm.1 NM_134161.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002842.mm.1 TC0800000371.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002843.mm.1 NM_001012517.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002843.mm.1 NM_001286422.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002843.mm.1 NM_001286424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002843.mm.1 NM_001286425.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002843.mm.1 NM_134161.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002843.mm.1 TC0800000371.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002839.mm.1 NM_001012517.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002839.mm.1 NM_001286422.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002839.mm.1 NM_001286425.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002839.mm.1 NM_134161.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002839.mm.1 TC0800000371.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002841.mm.1 NM_001012517.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002841.mm.1 NM_001286422.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002841.mm.1 NM_001286424.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002841.mm.1 NM_001286425.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002841.mm.1 NM_134161.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002841.mm.1 TC0800000371.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800002840.mm.1 NM_001286422.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800002840.mm.1 TC0800000371.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline