Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805430_m1
          See other MYPN GE Assays ›
          Gene Symbol:
          Mypn
          Gene Name:
          myopalladin
          Gene Aliases:
          1110056A04Rik, 9830112J03, AI853556, mKIAA4170
          Chromosome Location:
          Chr.10: 63115795 - 63203952 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805430_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mypn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182992.2 NP_892037.2 17 - 18 3716 68
          GenBank mRNA AK039718.1 - 1 - 2 787 68
          AK220521.1 - 17 - 18 3716 68
          BC052872.1 - 3 - 4 384 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002277.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mypn

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          sarcomere organization
          actin binding
          cytoskeletal protein binding
          SH3 domain binding
          muscle alpha-actinin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2706 BC052872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2706 AK004271.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2706 AK220521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2706 NM_182992.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2706 BC052872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2706 AK004271.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2706 AK220521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P2706 NM_182992.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857251 AK045753.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857251 BC032259.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857251 BC096463.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857251 NM_177565.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 705622 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243515 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652572 D28531.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45859 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284556 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948145 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 927972 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893522 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744344 AB023656.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744344 AF090190.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744344 AF131865.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744344 AK041278.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744344 AK147409.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744344 BC036964.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744344 BC059901.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744344 NM_207682.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931879 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665829 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 422246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 59465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 66499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814212 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127932 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701913 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 146883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82739 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 169605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 68908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 72555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 148573 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137985 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151350 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131216 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167970 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2706 BC052872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2706 AK004271.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2706 AK220521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2706 NM_182992.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2706 BC052872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2706 AK004271.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2706 AK220521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P2706 NM_182992.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167970 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131216 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151350 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07082278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07082155 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07082133 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137985 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07082254 - -
          Whole Mouse Genome G4122A A_52_P2706 BC052872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2706 AK004271.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2706 AK220521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2706 NM_182992.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2706 BC052872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2706 AK004271.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2706 AK220521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P2706 NM_182992.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291510 AK036458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291510 AK220521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P291510 NM_182992.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113901 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151350 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167970 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137985 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08731848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08603708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08256238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08529175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08868986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08814354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08674306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08320162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08473506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08557263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08256240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08414481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27189944 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10369718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369731 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369729 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369733 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369732 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369734 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369730 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369728 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369725 - -
          Affymetrix HT MG-430 PM Array Plate 1441551_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435813_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10369715 - -
          Mouse Genome 430 2.0 Array 1441551_at - -
          Mouse Genome 430 2.0 Array 1441551_PM_at - -
          Mouse Genome 430 2.0 Array 1435813_PM_at - -
          Mouse Genome 430 2.0 Array 1435813_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68802_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68802_at - -
          Affymetrix Mouse Clariom S Array PSR1000016336.mm.1 NM_182992.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016336.mm.1 NM_182992.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000016336.mm.1 TC1000002277.mm.1 MTA TC
          Mouse Expression Set 430 1435813_at - -
          Mouse Expression Set 430 1435813_PM_at - -
          Mouse Expression Set 430 1441551_at - -
          Mouse Expression Set 430 1441551_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441551_at - -
          Affymetrix HT Mouse Genome 430B Array 1435813_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441551_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435813_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 878738 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4540082 - -
          Illumina mouse-6 v1.0 expression beadchip 4540082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004540082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694687 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2719479 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694687 - -
          mouse-6 v1.1 (Illumina) scl37880.6.1_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 4540082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37880.6.1_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4540082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2694687 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2694687 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-xpdfw:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline