Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805460_m1
          See other ASIC3 GE Assays ›
          Gene Symbol:
          Asic3
          Gene Name:
          acid-sensing (proton-gated) ion channel 3
          Gene Aliases:
          AW742291, Accn3, DRASIC, SLNAC1, TNAC1
          Chromosome Location:
          Chr.5: 24410289 - 24417834 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805460_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Asic3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183000.2 NP_892045.2 1 - 2 792 82
          GenBank mRNA AY261387.1 - 1 - 2 531 82
          BC054460.1 - 1 - 2 792 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000214.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Asic3

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          cation transport
          sodium ion transport
          response to heat
          response to mechanical stimulus
          response to acidic pH
          ion transmembrane transport
          sodium ion transmembrane transport
          enterobactin transport
          detection of chemical stimulus involved in sensory perception
          sensory perception of sour taste
          detection of temperature stimulus involved in sensory perception
          detection of temperature stimulus involved in sensory perception of pain
          detection of mechanical stimulus involved in sensory perception of pain
          detection of chemical stimulus involved in sensory perception of pain
          detection of mechanical stimulus involved in sensory perception
          cation transmembrane transport
          cation channel activity
          sodium channel activity
          ligand-gated sodium channel activity
          PDZ domain binding
          enterobactin transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132496 AY261387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132496 AK028403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132496 BC054460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132496 NM_183000.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162727 BC054460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162727 NM_183000.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 663714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580551 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121036 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154505 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153308 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162727 BC054460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162727 NM_183000.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P609738 AY261387.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P609738 BC054460.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P609738 NM_183000.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P609738 AK028403.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23541639 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154505 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121036 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153308 - -
          Whole Mouse Genome G4122A A_52_P609738 AY261387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609738 BC054460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609738 NM_183000.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P609738 AK028403.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129195 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132496 AY261387.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132496 AK028403.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132496 BC054460.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132496 NM_183000.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121036 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153308 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154505 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675084 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675086 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675087 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675092 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04675105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23541651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04771957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04771950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04675092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04750053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04771967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04675087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04462405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04675104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04873907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04392530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04771958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04771955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04392531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04771956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04771965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23541674 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10520180 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520186 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520174 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520181 - -
          Affymetrix Mouse Gene 1.0 ST Array 10520179 - -
          Affymetrix HT MG-430 PM Array Plate 1439983_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10520173 - -
          Mouse Genome 430 2.0 Array 1439983_PM_a_at - -
          Mouse Genome 430 2.0 Array 1439983_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 171209_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 171209_at - -
          Affymetrix Mouse Clariom S Array PSR0500001584.mm.1 TC0500000214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001584.mm.1 NM_183000.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001584.mm.1 NM_183000.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001585.mm.1 TC0500000214.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500001585.mm.1 NM_183000.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500001585.mm.1 NM_183000.2 Refseq
          Mouse Expression Set 430 1439983_PM_a_at - -
          Mouse Expression Set 430 1439983_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1439983_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1439983_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 316711 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4200524 - -
          Illumina mouse-6 v1.0 expression beadchip 6940672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2615124 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699096 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2699096 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2615124 - -
          mouse-6 v1.1 (Illumina) scl0004047.1_78-S - -
          mouse-6 v1.1 (Illumina) scl0004191.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200524 - -
          Illumina Sentrix mouse-6.v1 bead array 6940672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0004191.1_1-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0004047.1_78-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940672 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2830611 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2615124 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2699096 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2699096 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2830611 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2615124 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline