Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805645_m1
          See other PRSS46 GE Assays ›
          Gene Symbol:
          Prss46
          Gene Name:
          protease, serine 46
          Gene Aliases:
          1700112C13Rik, 79H19B, Tessp6
          Chromosome Location:
          Chr.9: 110844506 - 110856522 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805645_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss46
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183103.2 NP_898926.2 3 - 4 574 122
          GenBank mRNA AK007173.1 - 3 - 4 573 122
          BC087878.1 - 2 - 3 531 30284660 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001473.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss46

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          molecular_function
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P397437 NM_183103.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P397437 AK007173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P397437 BC087878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P397437 NM_183103.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P397437 AK007173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P397437 BC087878.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 573092 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847689 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48410 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109100 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121067 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P397437 NM_183103.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P397437 AK007173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P397437 BC087878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P397437 NM_183103.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P397437 AK007173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P397437 BC087878.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26826214 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121067 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06926287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109100 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06926269 - -
          Whole Mouse Genome G4122A A_51_P397437 NM_183103.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397437 AK007173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397437 BC087878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397437 NM_183103.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397437 AK007173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P397437 BC087878.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118796 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109100 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121067 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08085982 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08090215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08085978 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08085981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08091147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07611254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08085244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07877863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08091148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07481756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07867738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07607826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08078423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08139423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08139424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08085976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07481757 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10589643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589642 - -
          Affymetrix HT MG-430 PM Array Plate 1429646_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10589640 - -
          Mouse Genome 430 2.0 Array 1429646_PM_at - -
          Mouse Genome 430 2.0 Array 1429646_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74306_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74306_at - -
          Affymetrix Mouse Clariom S Array PSR0900012332.mm.1 NM_183103.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900012332.mm.1 NM_183103.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900012332.mm.1 TC0900001473.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900012333.mm.1 NM_183103.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900012333.mm.1 NM_183103.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900012333.mm.1 TC0900001473.mm.1 MTA TC
          Mouse Expression Set 430 1429646_at - -
          Mouse Expression Set 430 1429646_PM_at - -
          Murine Genome U74v2 Set 168262_at - -
          Murine Genome U74v2 Set 168262_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429646_at - -
          Affymetrix HT Mouse Genome 430B Array 1429646_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 402422 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3130520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2625085 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2625085 - -
          mouse-6 v1.1 (Illumina) scl36418.5.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36418.5.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2625085 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2625085 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline