Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805670_g1
          See other 1700029I15RIK GE Assays ›
          Gene Symbol:
          1700029I15Rik
          Gene Name:
          RIKEN cDNA 1700029I15 gene
          Gene Aliases:
          1700021K07Rik
          Chromosome Location:
          Chr.2: 92382918 - 92383603 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805670_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700029I15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183112.3 NP_898935.1 2 - 3 215 67
          GenBank mRNA AK006499.1 - 2 - 3 216 67
          BC144843.1 - 2 - 3 278 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001412.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700029I15Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2047196 AK006214.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546660 BC094251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546660 NM_183112.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546660 AK006499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546660 BC094251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546660 NM_183112.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P546660 AK006499.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131475 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546660 BC094251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546660 NM_183112.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546660 AK006499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546660 BC094251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546660 NM_183112.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P546660 AK006499.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21428140 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131475 - -
          Whole Mouse Genome G4122A A_52_P546660 BC094251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P546660 NM_183112.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P546660 AK006499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P546660 BC094251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P546660 NM_183112.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P546660 AK006499.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107715 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131475 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01727384 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01727441 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01727398 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01727425 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01727388 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01727402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21428091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21428136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01727396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21428155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21428142 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474042 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474043 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474044 - -
          Affymetrix HT MG-430 PM Array Plate 1438499_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438979_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10474041 - -
          Mouse Genome 430 2.0 Array 1438979_PM_s_at - -
          Mouse Genome 430 2.0 Array 1438499_PM_at - -
          Mouse Genome 430 2.0 Array 1438499_at - -
          Mouse Genome 430 2.0 Array 1438979_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75641_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75641_at - -
          Affymetrix Mouse Clariom S Array PSR0200011046.mm.1 NM_183112.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011046.mm.1 NM_183112.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200011046.mm.1 TC0200001412.mm.1 MTA TC
          Mouse Expression Set 430 1438979_PM_s_at - -
          Mouse Expression Set 430 1438499_at - -
          Mouse Expression Set 430 1438979_s_at - -
          Mouse Expression Set 430 1438499_PM_at - -
          Murine Genome U74v2 Set 166361_f_at - -
          Murine Genome U74v2 Set 166362_at - -
          Murine Genome U74v2 Set 166361_PM_f_at - -
          Murine Genome U74v2 Set 166362_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438979_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1438979_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1438499_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438499_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 778163 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6840632 - -
          Illumina mouse-6 v1.0 expression beadchip 104060441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2632536 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2632536 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2469869 - -
          mouse-6 v1.1 (Illumina) scl20605.4.1_68-S - -
          mouse-6 v1.1 (Illumina) scl0075641.2_293-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840632 - -
          Illumina Sentrix mouse-6.v1 bead array 104060441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0075641.2_293-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104060441 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840632 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2632536 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2632536 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline