Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805727_m1
          See other SPINKL GE Assays ›
          Gene Symbol:
          Spinkl
          Gene Name:
          serine protease inhibitor, Kazal type-like
          Gene Aliases:
          9530002K18Rik
          Chromosome Location:
          Chr.18: 44166358 - 44176707 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805727_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spinkl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183123.2 NP_898946.1 4 - 5 271 145
          GenBank mRNA AK020535.1 - 4 - 5 272 145

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001278.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spinkl

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s)

          positive regulation of sperm motility
          negative regulation of sperm capacitation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972710 AK020535.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972710 EU234015.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972710 NM_183123.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162971 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565787 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 274708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 574945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 592437 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801197 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812852 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 959381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222671 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292188 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 416884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719971 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260227 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726738 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504845 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550424 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118368 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166644 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75309 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12394 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149117 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972710 AK020535.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972710 EU234015.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972710 NM_183123.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31665565 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149117 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08983708 - -
          Whole Mouse Genome G4122A A_51_P272113 AK020535.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133373 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133220 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14105971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14061794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14118755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13980212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31665555 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10458721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458724 - -
          Affymetrix HT MG-430 PM Array Plate 1455909_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10458719 - -
          Mouse Genome 430 2.0 Array 1455909_at - -
          Mouse Genome 430 2.0 Array 1455909_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77424_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77424_at - -
          Affymetrix Mouse Clariom S Array PSR1800009522.mm.1 NM_183123.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009522.mm.1 NM_183123.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009522.mm.1 TC1800001278.mm.1 MTA TC
          Mouse Expression Set 430 1455909_PM_at - -
          Mouse Expression Set 430 1455909_at - -
          Affymetrix HT Mouse Genome 430B Array 1455909_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455909_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 399160 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6380279 - -
          Illumina mouse-6 v1.0 expression beadchip 6380279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2627177 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2627177 - -
          mouse-6 v1.1 (Illumina) scl51452.5.1_124-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51452.5.1_124-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380279 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2627177 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline