Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805843_m1
          See other PLK5 GE Assays ›
          Gene Symbol:
          Plk5
          Gene Name:
          polo-like kinase 5
          Gene Aliases:
          6330514A18Rik
          Chromosome Location:
          Chr.10: 80356459 - 80365489 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805843_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plk5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183152.3 NP_898975.2 12 - 13 1471 120
          GenBank mRNA AK031968.1 - 12 - 13 1472 120
          BC086684.1 - 7 - 8 768 30679265 120
          BC099679.1 - 12 - 13 1440 6489647 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000853.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plk5

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response to tumor cell
          protein phosphorylation
          obsolete transformed cell apoptotic process
          cellular response to DNA damage stimulus
          cell cycle
          negative regulation of cell proliferation
          positive regulation of neuron projection development
          cell differentiation
          G2 DNA damage checkpoint
          cell division
          cellular response to growth factor stimulus
          regulation of G1/S transition of mitotic cell cycle
          nucleotide binding
          protein kinase activity
          ATP binding
          kinase activity
          transferase activity, transferring phosphorus-containing groups
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282702 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282701 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282703 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282704 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282699 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282700 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972297 BC086684.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972297 BC099679.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972297 NM_183152.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972297 AK031968.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 588979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166440 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112496 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972297 BC086684.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972297 BC099679.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972297 NM_183152.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972297 AK031968.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27282708 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167170 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112496 - -
          Whole Mouse Genome G4122A A_51_P328817 BC086684.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328817 BC099679.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328817 NM_183152.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328817 AK031968.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129654 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167170 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112496 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282703 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282702 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282700 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282699 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282704 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27282701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27282697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08499446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08794569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08207135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08588168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08588175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08207145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08588162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08904868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08499444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27282778 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364793 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364807 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364805 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364794 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364796 - -
          Affymetrix HT MG-430 PM Array Plate 1434984_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10364792 - -
          Mouse Genome 430 2.0 Array 1434984_at - -
          Mouse Genome 430 2.0 Array 1434984_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 216166_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 216166_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000006528.mm.1 NM_183152.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006528.mm.1 TC1000000853.mm.1 MTA TC
          Mouse Expression Set 430 1434984_PM_at - -
          Mouse Expression Set 430 1434984_at - -
          Murine Genome U74v2 Set 129082_at - -
          Murine Genome U74v2 Set 129082_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434984_at - -
          Affymetrix HT Mouse Genome 430B Array 1434984_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 480048 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5360435 - -
          Illumina mouse-6 v1.0 expression beadchip 5360435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360435 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216540 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216540 - -
          mouse-6 v1.1 (Illumina) scl38681.14.1_267-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38681.14.1_267-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360435 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl38681.14.1_267-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline