Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00805936_m1
          See other MPV17L2 GE Assays ›
          Gene Symbol:
          Mpv17l2
          Gene Name:
          MPV17 mitochondrial membrane protein-like 2
          Gene Aliases:
          Fksg24
          Chromosome Location:
          Chr.8: 70758649 - 70760921 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00805936_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mpv17l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183170.2 NP_898993.1 2 - 3 370 80
          GenBank mRNA AK138901.1 - 2 - 3 396 80
          AK151477.1 - 2 - 3 393 80
          AK152031.1 - 2 - 3 393 80
          AK168278.1 - 2 - 3 348 80
          AY013699.1 - 2 - 3 370 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002402.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mpv17l2

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial ribosome assembly
          positive regulation of mitochondrial translation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953857 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953851 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953852 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953853 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953856 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 AK152031.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 AK168278.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 AY013699.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 BC051227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 NM_183170.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 AK152031.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 AK168278.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 AY013699.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 BC051227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483311 NM_183170.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481283 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 AK152031.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 AK168278.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 AY013699.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 BC051227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 NM_183170.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 AK152031.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 AK168278.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 AY013699.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 BC051227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483311 NM_183170.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953857 - -
          Whole Mouse Genome G4122A A_51_P483311 AK152031.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483311 AK168278.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483311 AY013699.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483311 BC051227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483311 NM_183170.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483311 AK152031.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483311 AK168278.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483311 AY013699.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483311 BC051227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483311 NM_183170.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127694 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06950254 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06950244 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06950246 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953852 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953854 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953857 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953853 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06924612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06950248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06950245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06924598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06950244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953840 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10579343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579345 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579346 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579342 - -
          Affymetrix HT MG-430 PM Array Plate 1434006_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10579341 - -
          Mouse Genome 430 2.0 Array 1434006_PM_at - -
          Mouse Genome 430 2.0 Array 1434006_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234384_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 234384_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800018254.mm.1 NM_183170.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800018254.mm.1 NM_183170.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800018254.mm.1 TC0800002402.mm.1 MTA TC
          Mouse Expression Set 430 1434006_at - -
          Mouse Expression Set 430 1434006_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434006_at - -
          Affymetrix HT Mouse Genome 430B Array 1434006_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5900497 - -
          Illumina mouse-6 v1.0 expression beadchip 5900497 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005900497 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238114 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238114 - -
          mouse-6 v1.1 (Illumina) scl34694.5.9_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 5900497 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34694.5.9_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5900497 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238114 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238114 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl34694.5.9_23-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-dd86746ff-8rs9g:80/100.66.75.30:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline