Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fcrl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113238.1 | NP_001106709.1 | 2 - 3 | 184 | 70 | |
GenBank mRNA | AK089756.1 | - | 2 - 3 | 154 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |