Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00806364_m1
          See other 1700020N01RIK GE Assays ›
          Gene Symbol:
          1700020N01Rik
          Gene Name:
          RIKEN cDNA 1700020N01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 21593145 - 21622375 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00806364_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700020N01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027968.1 2 - 3 330 65
          GenBank mRNA AK006179.1 - 2 - 3 318 65
          AK007146.1 - 2 - 3 317 65
          BC099546.1 - 2 - 3 323 30920524 65
          CA464995.1 - 2 - 3 332 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000169.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071779 AK006179.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071779 AK007146.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071779 BC099546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071779 CA464995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071779 NR_027968.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718824 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 91427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106391 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970415 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137564 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46651 - -
          Whole Mouse Genome G4122A A_52_P662911 AK006179.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662911 AK007146.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662911 BC099546.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662911 CA464995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662911 NR_027968.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08553339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08718783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08588859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08865935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08588861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08274015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08355422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08488208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08588838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08444313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08427087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08553356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08274013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26993990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26994005 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10362067 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362068 - -
          Affymetrix HT MG-430 PM Array Plate 1429868_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10362064 - -
          Mouse Genome 430 2.0 Array 1429868_PM_at - -
          Mouse Genome 430 2.0 Array 1429868_at - -
          Affymetrix Mouse Clariom S Array PSR1000001223.mm.1 NR_027968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001223.mm.1 NR_027968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001223.mm.1 TC1000000169.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001225.mm.1 NR_027968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001225.mm.1 NR_027968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001225.mm.1 TC1000000169.mm.1 MTA TC
          Mouse Expression Set 430 1429868_at - -
          Mouse Expression Set 430 1429868_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429868_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429868_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 676829 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4810504 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594171 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2594171 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2795968 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2795968 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline