Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00806415_m1
          See other 4921517D22RIK GE Assays ›
          Gene Symbol:
          4921517D22Rik
          Gene Name:
          RIKEN cDNA 4921517D22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 59687402 - 59694103 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00806415_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4921517D22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183290.2 NP_899113.1 3 - 4 345 144
          GenBank mRNA AK014909.1 - 3 - 4 346 144
          AK133255.1 - 3 - 4 353 144
          BC147600.1 - 2 - 3 134 9056590 144
          BC147624.1 - 2 - 3 134 9056614 144

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002120.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4921517D22Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989757 AK133255.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989757 NM_183290.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14517 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566986 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141409 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162077 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989757 AK133255.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989757 NM_183290.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29081008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29080996 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07893562 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162077 - -
          Whole Mouse Genome G4122A A_52_P683553 BC147624.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P683553 AK133255.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P683553 BC147600.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P683553 NM_183290.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P683553 AK014909.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111967 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10629675 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10629680 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10629678 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10629674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10629677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29081047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29080974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10480226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29081018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29081001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10455469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10629682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29081020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11033529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10467300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29081015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10629672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10856648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29081022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10480234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29081008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11108182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29081006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29081004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29080946 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409784 - -
          Affymetrix HT MG-430 PM Array Plate 1430254_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409779 - -
          Mouse Genome 430 2.0 Array 1430254_PM_at - -
          Mouse Genome 430 2.0 Array 1430254_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70900_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70900_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300013599.mm.1 NM_183290.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013599.mm.1 NM_183290.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013599.mm.1 TC1300002120.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013600.mm.1 NM_183290.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013600.mm.1 NM_183290.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013600.mm.1 TC1300002120.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013601.mm.1 NM_183290.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013601.mm.1 NM_183290.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013601.mm.1 TC1300002120.mm.1 MTA TC
          Mouse Expression Set 430 1430254_PM_at - -
          Mouse Expression Set 430 1430254_at - -
          Murine Genome U74v2 Set 167659_PM_at - -
          Murine Genome U74v2 Set 167659_at - -
          Affymetrix HT Mouse Genome 430B Array 1430254_at - -
          Affymetrix HT Mouse Genome 430B Array 1430254_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 884180 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6900441 - -
          Illumina mouse-6 v1.0 expression beadchip 6900441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006900441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228780 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228780 - -
          mouse-6 v1.1 (Illumina) scl43878.5.1_241-S - -
          Illumina Sentrix mouse-6.v1 bead array 6900441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43878.5.1_241-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6900441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228780 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228780 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0