Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00806979_m1
          See other DEFB14 GE Assays ›
          Gene Symbol:
          Defb14
          Gene Name:
          defensin beta 14
          Gene Aliases:
          BD-14
          Chromosome Location:
          Chr.8: 19163156 - 19195309 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00806979_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defb14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183026.2 NP_898847.1 1 - 2 83 104
          GenBank mRNA AJ578468.2 - 1 - 2 54 104
          BC116199.1 - 1 - 2 54 40090056 104
          EH103347.1 - 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000179.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defb14

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          defense response to bacterium
          positive chemotaxis
          cell chemotaxis
          chemoattractant activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P68883 NM_183026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P68883 AJ578468.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P68883 BC116198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P68883 BC116199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P68883 NM_183026.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P68883 AJ578468.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P68883 BC116198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P68883 BC116199.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161846 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P68883 NM_183026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P68883 AJ578468.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P68883 BC116198.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P68883 BC116199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P68883 NM_183026.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P68883 AJ578468.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P68883 BC116198.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P68883 BC116199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P68883 NM_183026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P68883 AJ578468.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P68883 BC116198.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P68883 BC116199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P68883 NM_183026.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P68883 AJ578468.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P68883 BC116198.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P68883 BC116199.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139521 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06716673 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06716663 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06716668 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06716666 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07146228 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10570607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570608 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570606 - -
          Affymetrix Mouse Clariom S Array PSR0800001583.mm.1 NM_183026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001583.mm.1 NM_183026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001583.mm.1 TC0800000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001580.mm.1 NM_183026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001580.mm.1 NM_183026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001580.mm.1 TC0800000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001582.mm.1 NM_183026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001582.mm.1 NM_183026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001582.mm.1 TC0800000179.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800001581.mm.1 NM_183026.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001581.mm.1 NM_183026.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800001581.mm.1 TC0800000179.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161733 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161733 - -
          mouse-6 v1.1 (Illumina) GI_33942109-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161733 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161733 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline