Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00808138_m1
          See other NANOS3 GE Assays ›
          Gene Symbol:
          Nanos3
          Gene Name:
          nanos homolog 3 (Drosophila)
          Gene Aliases:
          nos3
          Chromosome Location:
          Chr.8: 84173730 - 84176945 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00808138_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nanos3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_194059.2 NP_918948.1 1 - 2 492 146
          GenBank mRNA AB095973.1 - 1 - 2 492 146
          BC119513.1 - 1 - 2 492 40055381 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002571.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nanos3

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          multicellular organism development
          germ cell development
          spermatogenesis
          negative regulation of translation
          cell differentiation
          oogenesis
          regulation of cell cycle
          positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          negative regulation of apoptotic signaling pathway
          RNA binding
          protein binding
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018641 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018640 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169804 NM_194059.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169804 AB095973.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71370 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169804 NM_194059.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169804 AB095973.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P105480 NM_194059.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P105480 AB095973.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P105480 BC119513.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P105480 BC119514.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26018583 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01783090 - -
          Whole Mouse Genome G4122A A_51_P105480 NM_194059.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105480 AB095973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105480 BC119513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105480 BC119514.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128714 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07037157 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07009755 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07009748 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07009746 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07112631 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07112621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07009753 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06941540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07037156 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018640 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018639 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26018641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26018578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06997375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07444234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07052051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07444236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07444230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07009758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06875913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07009746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26018590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07009755 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10580132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580134 - -
          Affymetrix Mouse Gene 1.1 ST Array 10580131 - -
          Affymetrix Mouse Clariom S Array PSR0800019621.mm.1 NM_194059.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019621.mm.1 NM_194059.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019621.mm.1 TC0800002571.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019620.mm.1 NM_194059.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019620.mm.1 NM_194059.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019620.mm.1 TC0800002571.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 774258 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4200204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720493 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720493 - -
          mouse-6 v1.1 (Illumina) scl0244551.2_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0244551.2_35-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2720493 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2720493 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline