Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00808218_g1
          See other ACTA1 GE Assays ›
          Gene Symbol:
          Acta1
          Gene Name:
          actin, alpha 1, skeletal muscle
          Gene Aliases:
          AA959943, Acta-2, Acts, Actsk-1
          Chromosome Location:
          Chr.8: 123891758 - 123894775 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00808218_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acta1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001272041.1 NP_001258970.1 4 - 5 635 134
          NM_009606.3 NP_033736.1 3 - 4 545 134
          GenBank mRNA AK027919.1 - 3 - 4 521 134
          BC014877.1 - 3 - 4 3587657 134
          M12866.1 - 3 - 4 499 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800003161.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acta1

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle thin filament assembly
          skeletal muscle fiber development
          nucleotide binding
          protein binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242187 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242184 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242181 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246854 BC014877.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P246854 BC014877.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760826 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127554 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246854 BC014877.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P246854 BC014877.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246854 BC014877.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P246854 BC014877.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26242194 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127554 - -
          Whole Mouse Genome G4122A A_51_P246854 BC014877.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P246854 BC014877.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132868 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127554 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07030304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06885188 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06885187 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06885184 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242187 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242184 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07146130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06842416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06881233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06881235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06881234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07194920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07160808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06839020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07030311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06842420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06881220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06839026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07030300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06927312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06839024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242198 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10582598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582597 - -
          Affymetrix HT MG-430 PM Array Plate 1427735_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10582592 - -
          Mouse Genome 430 2.0 Array 1427735_a_at - -
          Mouse Genome 430 2.0 Array 1427735_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11459_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11459_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800024252.mm.1 NM_001272041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024252.mm.1 NM_009606.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024252.mm.1 TC0800003161.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024251.mm.1 NM_001272041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024251.mm.1 NM_009606.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024251.mm.1 TC0800003161.mm.1 MTA TC
          Mouse Expression Set 430 1427735_a_at - -
          Mouse Expression Set 430 1427735_PM_a_at - -
          Murine Genome U74v2 Set 100381_at - -
          Murine Genome U74v2 Set 100381_PM_at - -
          Mouse Genome 430A 2.0 Array 1427735_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1427735_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427735_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427735_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 916274 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 840538 - -
          Illumina mouse-6 v1.0 expression beadchip 840538 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840538 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2738825 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2738825 - -
          mouse-6 v1.1 (Illumina) scl34188.7.1_72-S - -
          Illumina Sentrix mouse-6.v1 bead array 840538 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34188.7.1_72-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840538 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2738825 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2738825 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl34188.7.1_72-S - -

          Back To Top
           

          Citations (1)

          Stimulation of skeletal muscle myofibrillar protein synthesis, p70 S6 kinase phosphorylation, and ribosomal protein S6 phosphorylation by inhibition of myostatin in mature mice.
          Authors
          Welle S, Burgess K, Mehta S
          Journal
          Am J Physiol Endocrinol Metab - Page No E567 (Sun Mar 01 00:00:00 UTC 2009)
          Volume (Year)
          296 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline