Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00809004_s1
          See other KRTAP13-1 GE Assays ›
          Gene Symbol:
          Krtap13-1
          Gene Name:
          keratin associated protein 13-1
          Gene Aliases:
          AA589563, KAP13.1
          Chromosome Location:
          Chr.16: 88728862 - 88729611 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00809004_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krtap13-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183189.1 NP_899012.1 1 678 113
          GenBank mRNA BC027551.1 - 1 678 1363815 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krtap13-1

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P135118 NM_183189.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P135118 BC027551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P135118 NM_183189.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P135118 BC027551.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139309 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P135118 NM_183189.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P135118 BC027551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P135118 NM_183189.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P135118 BC027551.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30947632 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139309 - -
          Whole Mouse Genome G4122A A_51_P135118 NM_183189.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135118 BC027551.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135118 NM_183189.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P135118 BC027551.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100028 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139309 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13012436 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13012442 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13012452 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13012453 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13012433 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13012451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30947647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13012455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12941690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12898611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12992918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12992917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12853608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12992915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12891825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12486571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12477962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12734392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12720915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12720918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12980382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12743728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12853610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12734390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12486586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30947556 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10436747 - -
          Affymetrix HT MG-430 PM Array Plate 1428007_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10436746 - -
          Mouse Genome 430 2.0 Array 1428007_at - -
          Mouse Genome 430 2.0 Array 1428007_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 268905_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 268905_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600007556.mm.1 NM_183189.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007556.mm.1 NM_183189.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007556.mm.1 TC1600000963.mm.1 MTA TC
          Mouse Expression Set 430 1428007_PM_at - -
          Mouse Expression Set 430 1428007_at - -
          Mouse Genome 430A 2.0 Array 1428007_PM_at - -
          Mouse Genome 430A 2.0 Array 1428007_at - -
          Affymetrix HT Mouse Genome 430A Array 1428007_at - -
          Affymetrix HT Mouse Genome 430A Array 1428007_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 801889 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7040438 - -
          Illumina mouse-6 v1.0 expression beadchip 7040438 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040438 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2657135 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2657135 - -
          mouse-6 v1.1 (Illumina) scl48904.1.23_262-S - -
          Illumina Sentrix mouse-6.v1 bead array 7040438 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48904.1.23_262-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040438 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2657135 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2657135 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline