Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00809442_s1
          See other FFAR1 GE Assays ›
          Gene Symbol:
          Ffar1
          Gene Name:
          free fatty acid receptor 1
          Gene Aliases:
          Gpr40
          Chromosome Location:
          Chr.7: 30860568 - 30861470 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00809442_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ffar1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_194057.2 NP_918946.2 1 854 147
          GenBank mRNA AB095745.1 - 1 854 147
          BC132014.1 - 1 948 40130291 147
          BC138099.1 - 1 948 8861118 147

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ffar1

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          positive regulation of calcium ion transport into cytosol
          insulin secretion
          positive regulation of insulin secretion
          glucose homeostasis
          positive regulation of calcium ion transport
          response to fatty acid
          signal transducer activity
          G-protein coupled receptor activity
          fatty acid binding
          lipid binding
          bioactive lipid receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P551974 AB095745.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P551974 BC132014.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P551974 BC138099.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P551974 NM_194057.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P551974 AB095745.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P551974 BC132014.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P551974 BC138099.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P551974 NM_194057.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154161 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P551974 AB095745.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P551974 BC132014.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P551974 BC138099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P551974 NM_194057.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P551974 AB095745.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P551974 BC132014.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P551974 BC138099.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P551974 NM_194057.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033788 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154161 - -
          Whole Mouse Genome G4122A A_52_P551974 AB095745.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P551974 BC132014.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P551974 BC138099.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P551974 NM_194057.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P551974 AB095745.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P551974 BC132014.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P551974 BC138099.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P551974 NM_194057.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130822 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154161 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05980825 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06074666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06074673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06074676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06332150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06024065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06660410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06332156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562131 - -
          Affymetrix Mouse Gene 1.1 ST Array 10562130 - -
          Affymetrix Mouse Clariom S Array PSR0700024797.mm.1 TC0700002778.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024799.mm.1 TC0700002778.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024798.mm.1 TC0700002778.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024798.mm.1 NM_194057.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024798.mm.1 NM_194057.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 540112 AK086362.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 540112 AK157156.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 540112 BC059886.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 540112 NM_172566.2 refseq_mrna_mouse_NM
          Illumina mouseRef-8 v1.0 expression beadchip 0000540112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2750208 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2750208 - -
          mouse-6 v1.1 (Illumina) scl0233081.1_313-S - -
          Illumina Sentrix mouse-6.v1 bead array 540112 AK086362.1 genbank_mrna_mouse
          Illumina Sentrix mouse-6.v1 bead array 540112 AK157156.1 genbank_mrna_mouse
          Illumina Sentrix mouse-6.v1 bead array 540112 BC059886.1 genbank_mrna_mouse
          Illumina Sentrix mouse-6.v1 bead array 540112 NM_172566.2 refseq_mrna_mouse_NM
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0233081.1_313-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540112 AK086362.1 genbank_mrna_mouse
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540112 AK157156.1 genbank_mrna_mouse
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540112 BC059886.1 genbank_mrna_mouse
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540112 NM_172566.2 refseq_mrna_mouse_NM
          MouseRef-8 v2 Expression BeadChip ILMN_2750208 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2750208 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline