Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00810549_s1
          See other ADAM30 GE Assays ›
          Gene Symbol:
          Adam30
          Gene Name:
          a disintegrin and metallopeptidase domain 30
          Gene Aliases:
          4933424D07Rik, svph4
          Chromosome Location:
          Chr.3: 98160812 - 98163173 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00810549_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adam30
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027665.1 NP_081941.1 1 2143 126
          GenBank mRNA AK077058.1 - 2296 126
          AY190760.1 - 1 2143 126
          BC113152.1 - 1 2135 40059344 126
          BC113762.1 - 1 2135 40059342 126

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adam30

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 BC113762.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 AY190760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 BC113152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 NM_027665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 AK016888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 BC113762.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 AY190760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 BC113152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 NM_027665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P539250 AK016888.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460905 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 3948 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156331 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 BC113762.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 AY190760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 BC113152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 NM_027665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 AK016888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 BC113762.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 AY190760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 BC113152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 NM_027665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P539250 AK016888.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P202305 AK077058.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22358765 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156331 - -
          Whole Mouse Genome G4122A A_52_P539250 BC113762.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P539250 AY190760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P539250 BC113152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P539250 NM_027665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P539250 AK016888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P539250 BC113762.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P539250 AY190760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P539250 BC113152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P539250 NM_027665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P539250 AK016888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P202305 AK077058.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124076 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156331 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02653223 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02331487 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02843997 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02843999 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02331481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22358717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02331501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03164867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02653223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02331486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02568787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02331500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02331498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03005841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02331483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02767869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03063817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22358715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02576156 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10494634 - -
          Affymetrix Mouse Gene 1.1 ST Array 10494632 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 420883 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 520037 - -
          MouseWG-6 v2 Expression BeadChip 104540181 - -
          Illumina mouse-6 v1.0 expression beadchip 520037 - -
          Illumina mouse-6 v1.0 expression beadchip 104540181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2763590 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2439745 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2763590 - -
          mouse-6 v1.1 (Illumina) scl0071078.2_199-S - -
          mouse-6 v1.1 (Illumina) scl22835.1_109-S - -
          Illumina Sentrix mouse-6.v1 bead array 520037 - -
          Illumina Sentrix mouse-6.v1 bead array 104540181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0071078.2_199-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104540181 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2890895 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2439745 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2763590 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2890895 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0071078.2_199-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline