Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00812850_g1
          See other ADM2 GE Assays ›
          Gene Symbol:
          Adm2
          Gene Name:
          adrenomedullin 2
          Gene Aliases:
          Am2, IMD, Imdn
          Chromosome Location:
          Chr.15: 89322899 - 89324731 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00812850_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adm2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182928.4 NP_891558.1 2 - 3 359 92
          GenBank mRNA AB121035.1 - 2 - 3 283 92
          AF529212.1 - 1 - 2 113 92
          BC096428.1 - 2 - 3 286 4948766 92
          CJ052708.1 - 2 - 3 360 92
          DV649647.1 - 2 - 3 235 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000869.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adm2

          Panther Classification:

          Molecular Function -

          peptide hormone intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          protein phosphorylation
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          digestion
          feeding behavior
          positive regulation of gene expression
          positive regulation of angiogenesis
          negative regulation of blood pressure
          hormone activity
          protein complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424509 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181386 BC096428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181386 NM_182928.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181386 AB121035.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321136 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117698 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181386 BC096428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181386 NM_182928.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181386 AB121035.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424540 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117698 - -
          Whole Mouse Genome G4122A A_52_P458169 BC096428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458169 AF529212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458169 NM_182928.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P458169 AB121035.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121930 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117698 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12423112 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424514 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424512 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424513 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424515 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424510 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424516 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30424507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12423127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12036748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12423110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30424547 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10426209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426211 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426212 - -
          Affymetrix HT MG-430 PM Array Plate 1442856_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10426208 - -
          Mouse Genome 430 2.0 Array 1442856_PM_at - -
          Mouse Genome 430 2.0 Array 1442856_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223780_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223780_at - -
          Affymetrix Mouse Clariom S Array PSR1500006823.mm.1 NM_182928.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006823.mm.1 TC1500000869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006824.mm.1 NM_182928.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006824.mm.1 TC1500000869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006826.mm.1 NM_182928.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006826.mm.1 TC1500000869.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500006825.mm.1 NM_182928.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1500006825.mm.1 TC1500000869.mm.1 MTA TC
          Mouse Expression Set 430 1442856_at - -
          Mouse Expression Set 430 1442856_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442856_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442856_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 795418 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4060102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241813 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241813 - -
          mouse-6 v1.1 (Illumina) scl47621.3.1_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060102 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47621.3.1_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1241813 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241813 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline