| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100012846.mm.1 |
NM_001172099.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012846.mm.1 |
NM_001172099.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012846.mm.1 |
NM_198013.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012846.mm.1 |
NM_198013.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012846.mm.1 |
TC1100001363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012848.mm.1 |
NM_001172099.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012848.mm.1 |
NM_001172099.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012848.mm.1 |
NM_198013.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012848.mm.1 |
NM_198013.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012848.mm.1 |
TC1100001363.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100012847.mm.1 |
NM_001172099.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012847.mm.1 |
NM_001172099.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012847.mm.1 |
NM_198013.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012847.mm.1 |
NM_198013.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100012847.mm.1 |
TC1100001363.mm.1 |
MTA TC |